Skip to Content
Merck
All Photos(1)

Key Documents

139513

Sigma-Aldrich

1-Methyl-2-pyrrolidine ethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.4713 (lit.)

bp

110-112 °C/14 mmHg (lit.)

density

0.951 g/mL at 25 °C (lit.)

SMILES string

CN1CCCC1CCO

InChI

1S/C7H15NO/c1-8-5-2-3-7(8)4-6-9/h7,9H,2-6H2,1H3

InChI key

FYVMBPXFPFAECB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methyl-2-pyrrolidine ethanol is an important pharmaceutical intermediate.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrogenation of pyrrole derivatives I. Hydrogenations over palladium.
Mathe T and Tungler A.
Applied Catalysis A: General, 143(2), 309-316 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service