GF41621183
Inconel® X750 - heat resisting alloy, Ni74Cr15Fe7TiAlNb
foil, 0.5m coil, 0.25mm thickness, annealed
Synonym(s):
NI160270
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
Recommended Products
form
annealed
foil (coil)
manufacturer/tradename
Goodfellow 416-211-83
size × thickness
0.5 m × 0.25 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service