Skip to Content
Merck
All Photos(3)

Key Documents

428043

Sigma-Aldrich

Polyethylene

low density, melt index 25 g/10 min (190°C/2.16kg)

Synonym(s):

PE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

pellets

melt index

25 g/10 min (190°C/2.16kg)

impact strength

45.4 J/m (Izod, ASTM D 256, −50 °C, notched)

mp

116 °C

transition temp

softening point 93 °C (Vicat, ASTM D 1525)

density

0.925 g/mL at 25 °C

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Injection and blow molded toys, housewares and lids.

Features and Benefits

Good toughness, processability and appearance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flora Rendell-Bhatti et al.
Environmental pollution (Barking, Essex : 1987), 269, 115744-115744 (2020-12-02)
Microplastic pollution has become ubiquitous, affecting a wide variety of biota. Although microplastics are known to alter the development of a range of marine invertebrates, no studies provide a detailed morphological characterisation of the developmental defects. Likewise, the developmental toxicity
Lisa K McCarthy et al.
Pediatrics, 132(1), e135-e141 (2013-06-19)
Hypothermia on admission to the NICU is associated with increased mortality in preterm infants. Many newborns are hypothermic on admission despite using polyethylene bags (PBs). Using exothermic mattresses (EMs) in addition to PBs may reduce hypothermia but increase hyperthermia. We
Łukasz Krakowxzyk et al.
Medical science monitor basic research, 19, 210-213 (2013-08-22)
The search for new surgical flap techniques and modifications of already existing ones is gaining increasing popularity. Progress in flap designing and harvesting have improved the functional and aesthetic results, especially in head and neck reconstruction. Ten pigs were used
Katharine S Woods et al.
American journal of veterinary research, 74(4), 523-529 (2013-03-28)
To compare the diagnostic quality of bronchoalveolar lavage (BAL) fluid acquired from healthy dogs by manual aspiration via polyethylene tubing (MAPT) and via suction pump aspiration (SPA) with a suction trap connection. 12 healthy adult Beagles. BAL was performed with
Agnieszka Karbownik et al.
Acta poloniae pharmaceutica, 70(2), 301-307 (2013-04-26)
The study concerned the stability of calcium folinate in concentrate in glass vials and diluted in polyethylene (PE) bags stored at 15-25 degrees C and 2-8 degrees C for up to 34 days. Original vials of calcium folinate injection (10

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the average molecular weight of this polymer?

    1 answer
    1. The molecular weight of this item is not determined.

      Helpful?

  4. You didn't point the size, Please write The particle sizes

    1 answer
    1. The particle size is not determined. In general, pellets can range from 2-6 mm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service