E14659
2-Ethyl-1-butanol
98%
Synonym(s):
2-Ethylbutyl alcohol, ‘Isohexylalcohol’
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(C2H5)2CHCH2OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731254
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.422 (lit.)
bp
146 °C (lit.)
mp
−15 °C (lit.)
density
0.83 g/mL at 25 °C (lit.)
SMILES string
CCC(CC)CO
InChI
1S/C6H14O/c1-3-6(4-2)5-7/h6-7H,3-5H2,1-2H3
InChI key
TZYRSLHNPKPEFV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
136.4 °F - closed cup
Flash Point(C)
58 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mar Serrano-Arnaldos et al.
Journal of biotechnology, 325, 91-99 (2020-11-15)
Esters from branched alcohols and dicarboxylic linear acids are widely used as lube bases due to their good performance at low temperatures. This work proposes a new process to synthesize bis(2-ethylbutyl) adipate and bis(2-ethylbutyl) sebacate by using the lipase-based catalyst
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service