Skip to Content
Merck
All Photos(2)

Key Documents

183105

Sigma-Aldrich

3,3-Dimethyl-1-butanol

97%

Synonym(s):

Neohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH2OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731466
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

143 °C (lit.)

mp

−60 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(C)CCO

InChI

1S/C6H14O/c1-6(2,3)4-5-7/h7H,4-5H2,1-3H3

InChI key

DUXCSEISVMREAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,3-Dimethyl-1-butanol is a glass forming material. The molecular dynamics of 3,3-dimethyl-1-butanol was studied.

Application

3,3-Dimethyl-1-butanol is a branched alkanol, which can be used:
  • As a ligand in the preparation of nickel based single-molecule magnets.
  • In the synthesis of lipophilic alkyl parabens as antifungal agents.
  • As a precursor to synthesize hydrocarbon surfactants.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast magnetization tunneling in tetranickel (II) single-molecule magnets.
Yang E-C, et al.
Inorganic Chemistry, 45(2), 529-546 (2006)
Thermal and relaxational properties of glass-forming material, 3, 3-dimethyl-1-butanol.
The Journal of Chemical Thermodynamics, 36(10), 877-888 (2004)
P Barlow et al.
Biochemical pharmacology, 34(17), 3117-3122 (1985-09-01)
3,3-Dimethylbutanol (Dimbunol), a competitive inhibitor of choline dehydrogenase (CDH), and ethylcholine mustard aziridinium (ECMA), an effective irreversible inhibitor of both CDH and choline transport, were investigated for their effects upon the uptake and metabolism of [3H]choline in mice. Thirty minutes
R M Krupka et al.
The Journal of biological chemistry, 255(18), 8546-8549 (1980-09-25)
Half-saturation constants have been determined for the choline carrier with cationic substrates and their uncharged carbon analogs: (a) choline and 3,3-dimethyl-1-butanol and (b) 2-dimethylaminoethanol and isoamyl alcohol. The constants are 6.3 microM and 16 mM for the first pair, and
Single-molecule magnets: high-field electron paramagnetic resonance evaluation of the single-ion zero-field interaction in a ZnII3NiII complex.
Yang E-C, et al.
Inorganic Chemistry, 44(11), 3827-3836 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service