Skip to Content
Merck
All Photos(3)

Key Documents

D1807

Sigma-Aldrich

1-Decene

94%

Synonym(s):

1-n-Decene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH2
CAS Number:
Molecular Weight:
140.27
Beilstein:
1737236
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.84 (vs air)

vapor pressure

1.6 mmHg ( 23.8 °C)

Assay

94%

form

liquid

autoignition temp.

455 °F

impurities

≤2.0% 2-butyl-1-hexene
≤2.0% 2-ethyl-1-octene
≤2.0% n-decane

refractive index

n20/D 1.421 (lit.)

bp

166.5-173.5 °C (lit.)

mp

−66.3-−66 °C (lit.)

density

0.741 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC=C

InChI

1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3

InChI key

AFFLGGQVNFXPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Decene is a linear alpha-olefin monomer used to produce poly-alpha-olefins (PAOs), which are crucial in various industrial applications, including lubricant formulation and as a co-monomer in the production of specialty polymers.

Application

  • Polymerization Catalyst Research with 1-Decene: A study on high molecular weight copolymers incorporating 1-Decene highlights advanced polymerization techniques using Lewis acid catalysts, essential for producing materials with specific mechanical properties (Wan et al., 2024).
  • 1-Decene in Biodegradable Polymer Development: Research into the molecular dynamics of 1-Decene oligomers assesses their suitability in the synthesis of high-performance, biodegradable polymers, contributing to more sustainable material solutions (Mercier Franco and Firoozabadi, 2023).
  • 1-Decene as a Precursor in Surfactant Production: An examination of the thermophysical properties of long linear and branched alkanes, including 1-Decene, provides insights into their potential as precursors for surfactant production, crucial for chemical engineering applications (Schmitt et al., 2023).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Computation of shear viscosity by a consistent method in equilibrium molecular dynamics simulations: applications to 1-decene oligomers
Mercier Franco, Lui?s Fernando and Firoozabadi, Abbas
The Journal of Physical Chemistry B, 127, 10043-10051 (2023)
Poly-$\alpha$-olefin-based synthetic lubricants: a short review on various synthetic routes
Ray, Saptarshi and Rao, et, al.
Lubrication Science, 24, 23-44 (2012)
Rong Shen et al.
PLoS computational biology, 11(10), e1004368-e1004368 (2015-10-28)
The knowledge of multiple conformational states is a prerequisite to understand the function of membrane transport proteins. Unfortunately, the determination of detailed atomic structures for all these functionally important conformational states with conventional high-resolution approaches is often difficult and unsuccessful.
Ilya Borisov et al.
Polymers, 12(6) (2020-05-30)
For the first time, the effect of the side-chain in polyalkylmethylsiloxane towards pervaporative removal of methyl tert-butyl ether (MTBE) from water was studied. The noticeable enhancement of separation factor during the pervaporation of 1 wt.% MTBE solution in water through
Antonios Pantazis et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(51), 18381-18386 (2014-12-10)
Excitation-evoked Ca(2+) influx is the fastest and most ubiquitous chemical trigger for cellular processes, including neurotransmitter release, muscle contraction, and gene expression. The voltage dependence and timing of Ca(2+) entry are thought to be functions of voltage-gated calcium (CaV) channels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service