Merck
  • High Selective Composite Polyalkylmethylsiloxane Membranes for Pervaporative Removal of MTBE from Water: Effect of Polymer Side-chain.
High Selective Composite Polyalkylmethylsiloxane Membranes for Pervaporative Removal of MTBE from Water: Effect of Polymer Side-chain.
Polymers (2020-05-30)
Ilya Borisov, Ivan Podtynnikov, Evgenia Grushevenko, Olga Scharova, Tatiana Anokhina, Sergey Makaev, Alexey Volkov, Vladimir Volkov
ABSTRACT
For the first time, the effect of the side-chain in polyalkylmethylsiloxane towards pervaporative removal of methyl tert-butyl ether (MTBE) from water was studied. The noticeable enhancement of separation factor during the pervaporation of 1 wt.% MTBE solution in water through the dense film (40-50 µm) can be achieved by substitution of a methyl group (separation factor 111) for heptyl (161), octyl (169) or decyl (180) one in polyalkylmethylsiloxane. Composite membrane with the selective layer (~8 µm) made of polydecylmethylsiloxane (M10) on top of microfiltration support (MFFK membrane) demonstrated MTBE/water separation factor of 310, which was 72% greater than for the dense film (180). A high separation factor together with an overall flux of 0.82 kg·m-2·h-1 allowed this M10/MFFK composite membrane to outperform the commercial composite membranes. The analysis of the concentration polarization modulus and the boundary layer thickness revealed that the feed flow velocity should be gradually increased from 5 cm·s-1 for an initial solution (1 wt.% of MTBE in water) to 13 cm·s-1 for a depleted solution (0.2 wt.% of MTBE in water) to overcome the concentration polarization phenomena in case of composite membrane M10/MFFK (Texp = 50 °C).
Sigma-Aldrich
1,3-Divinyltetramethyldisiloxane, 97%
Sigma-Aldrich
1-Decene, 94%
Sigma-Aldrich
1,7-Octadiene, 98%
Sigma-Aldrich
1-Heptene, 97%