All Photos(1)
About This Item
Linear Formula:
CH3(CH2)4CH=CH2
CAS Number:
Molecular Weight:
98.19
Beilstein:
1098332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
101 mmHg ( 37.7 °C)
Assay
≥99%
form
liquid
autoignition temp.
505 °F
refractive index
n20/D 1.400 (lit.)
bp
94 °C (lit.)
mp
−119 °C (lit.)
density
0.697 g/mL at 25 °C (lit.)
SMILES string
CCCCCC=C
InChI
1S/C7H14/c1-3-5-7-6-4-2/h3H,1,4-7H2,2H3
InChI key
ZGEGCLOFRBLKSE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
15.8 °F - closed cup
Flash Point(C)
-9.0 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hiroshi Toda et al.
Applied and environmental microbiology, 81(6), 1919-1925 (2015-01-04)
We describe the development of biocatalysis for producing optically pure straight-chain (S)-epoxyalkanes using styrene monooxygenase of Rhodococcus sp. strain ST-10 (RhSMO). RhSMO was expressed in the organic solvent-tolerant microorganism Kocuria rhizophila DC2201, and the bioconversion reaction was performed in an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service