Skip to Content
Merck
All Photos(2)

Key Documents

A7256

Sigma-Aldrich

DL-Norepinephrine hydrochloride

crystalline, ≥97% (TLC)

Synonym(s):

(±)-1-(3,4-Dihydroxyphenyl)-2-aminoethanol hydrochloride, (±)-4-(2-Amino-1-hydroxyethyl)-1,2-benzenediol hydrochloride, DL-Arterenol hydrochloride, DL-Noradrenaline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH(OH)CH2NH2 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3707003
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97% (TLC)

form

crystalline

color

off-white to tan

solubility

water: soluble 50 mg/mL, clear to slightly hazy, faintly yellow to brownish-yellow

storage temp.

−20°C

SMILES string

NCC(C1=CC(O)=C(C=C1)O)O.Cl

InChI

1S/C8H11NO3.ClH/c9-4-8(12)5-1-2-6(10)7(11)3-5;/h1-3,8,10-12H,4,9H2;1H

InChI key

FQTFHMSZCSUVEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Norepinephrine hydrochloride is an adrenergic drug.

Application

DL-Norepinephrine hydrochloride is suitable adrenergic drug used in the following studies:
  • To investigate the effects of adrenergic drugs on lantern of larval fly.
  • To evaluate the ability of α-adrenergic stimulation in the presence of elevated calcium levels to induce delayed after depolarizations and triggered activity in Purkinje fibers isolated from cat hearts.
  • Simultaneous reduction and surface functionalization of graphene oxide by a one-step poly(norepinephrine) functionalization.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous Reduction and Surface Functionalization of Graphene Oxide by Mussel-Inspired Chemistry.
Kang SM, et al.
Advances in Functional Materials, 21(1), 108-112 (2011)
S Kimura et al.
Circulation, 70(6), 1074-1082 (1984-12-01)
We studied the ability of alpha-adrenergic stimulation to induce delayed afterdepolarizations and triggered activity in Purkinje fibers from cat hearts in the presence of an elevated Ca++ concentration. Delayed afterdepolarizations could not be induced at drive cycle lengths of 200
Yining Chen et al.
Journal of neurophysiology, 125(6), 2397-2407 (2021-05-13)
Sensory learning during critical periods in development has lasting effects on behavior. Neuromodulators like dopamine and norepinephrine (NE) have been implicated in various forms of sensory learning, but little is known about their contribution to sensory learning during critical periods.
Effect of adrenergic drugs on the lantern of the larval Photuris firefly.
A D Carlson
The Journal of experimental biology, 48(2), 381-387 (1968-04-01)
Alexis N Orr et al.
International journal of molecular sciences, 22(5) (2021-04-04)
Transglutaminases (TGs) are crosslinking enzymes best known for their vascular remodeling in hypertension. They require calcium to form an isopeptide bond, connecting a glutamine to a protein bound lysine residue or a free amine donor such as norepinephrine (NE) or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service