Skip to Content
Merck
All Photos(4)

Key Documents

A9512

Sigma-Aldrich

L-(−)-Norepinephrine (+)-bitartrate salt monohydrate

≥99% (TLC), solid, vasoconstrictor

Synonym(s):

L-(−)-Noradrenaline (+)-bitartrate salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid

Assay

≥99%

form

solid

color

white

solubility

H2O: 46 mg/mL

SMILES string

O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.NC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C8H11NO3.C4H6O6.H2O/c9-4-8(12)5-1-2-6(10)7(11)3-5;5-1(3(7)8)2(6)4(9)10;/h1-3,8,10-12H,4,9H2;1-2,5-6H,(H,7,8)(H,9,10);1H2/t8-;1-,2-;/m01./s1

InChI key

LNBCGLZYLJMGKP-LUDZCAPTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-(-)-Norepinephrine (+)-bitartrate salt monohydrate has been used:
  • to infuse dogs to form a intracellular lipid accumulation model[1]
  • to analyse the signaling involved in the up-regulation of monocarboxylate transporter 2 (MCT2) expression in cultured cortical neuron[2]
  • in neurochemical preparation to study the response of gram-negative and gram-positive bacteria to norepinephrine[3]
  • to study the roles of peripheral hormones on thyroid activity of turtles[4]

Biochem/physiol Actions

Norepinephrine bitartrate plays an important role in in vivo and in vitro development of pathogenic and indigenous bacteria.[3] It acts as a vasoconstrictor and adrenergic neurotransmitter.

Caution

Light sensitive

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roles of thyroid, adrenal and pancreatic hormones on thyroid activity of the soft-shelled turtles Lissemys punctata punctata Bonnoterre
Ray PP, et al.
Folia Biologica (Krakow.), 54(3-4), 93-102 (2006)
Noradrenaline enhances the expression of the neuronal monocarboxylate transporter MCT2 by translational activation via stimulation of PI3K/Akt and the mTOR/S6K pathway
Chenal J and Pellerin L
Journal of Neurochemistry, 102(2), 389-397 (2007)
Jonathan E Wingo et al.
Journal of applied physiology (Bethesda, Md. : 1985), 109(5), 1301-1306 (2010-08-14)
Sweat rate (SR) is reduced in locally cooled skin, which may result from decreased temperature and/or parallel reductions in skin blood flow. The purpose of this study was to test the hypotheses that decreased skin blood flow and decreased local
Measurement of intracellular triglyceride stores by H spectroscopy: validation in vivo
Szczepaniak LS, et al.
American Journal of Physiology. Endocrinology and Metabolism, 276(5), E977-E989 (1999)
Stimulation of bacterial growth by heat-stable, norepinephrine-induced autoinducers
Freestone PPE, et al.
FEMS Microbiology Letters, 172(1), 53-60 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service