Skip to Content
Merck

Skip To

A7257

(−)-Norepinephrine

≥98% (TLC), crystalline, adrenergic neurotransmitter

Synonym(s):

(R)-4-(2-Amino-1-hydroxyethyl)-1,2-benzenediol, L-Arterenol, L-Noradrenaline, Levarterenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H11NO3
CAS Number:
Molecular Weight:
169.18
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-096-6
MDL number:
Beilstein/REAXYS Number:
2804840
Assay:
≥98%
Form:
crystalline
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

(−)-Norepinephrine, ≥98%, crystalline

Quality Segment

assay

≥98%

form

crystalline

color

off-white to tan

storage temp.

−20°C

SMILES string

NC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C8H11NO3/c9-4-8(12)5-1-2-6(10)7(11)3-5/h1-3,8,10-12H,4,9H2/t8-/m0/s1

InChI key

SFLSHLFXELFNJZ-QMMMGPOBSA-N

General description

Norepinephrine is a neurotransmitter found in both the peripheral and central nervous systems. It triggers effects associated with the ′fight or flight′ response and is mediated by the family of adrenergic receptors. Structurally, it is a catecholamine due to the presence of the catechol moiety (consisting of two hydroxyl groups on a benzene ring) and an amine (NH2) group. Norepinephrine exhibits pharmacologic effects on α1 and β1 adrenoceptors. It does not elevate heart rate. The primary advantage of norepinephrine is to improve organ perfusion by increasing vascular tone. Compared to dopamine, norepinephrine is more effective at achieving targeted endpoints, less metabolically active than epinephrine, and reduces serum lactate levels. Moreover, it notably enhances renal perfusion and splanchnic blood flow in sepsis, especially when used alongside dobutamine. Norepinephrine is used in the treatment of hypotension, depression, schizophrenia, and attention deficit/hyperactivity disorders.

Application

(−)-Norepinephrine has been used:
  • as anαand β-AR agonist to stimulate mouse organoids to study the pathways that are activated upon intestinal epithelial adrenergic receptor (AR) stimulation,
  • to measure the monoamine levels in various limbic regions of mouse brains by high-performance liquid chromatography (HPLC) coupled with an electrochemical equipped with an auto-sampler,
  • as a dispersal agent in biofilm dispersion assay to study its effects on the dispersal of Mannheimia haemolytica biofilms

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E4250N5785E4375
description

≥98%, crystalline

description

-

description

meets USP testing specifications

description

solid

assay

≥98%

assay

-

assay

-

assay

-

form

crystalline

form

-

form

solid

form

solid

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

color

off-white to tan

color

-

color

-

color

white

Gene Information

human ... ADRA1A(148), ADRA1B(147), ADRA1D(146), ADRA2A(150), ADRA2B(151), ADRA2C(152), ADRB1(153), ADRB2(154), ADRB3(155)
rat ... Adra1a(29412), Adra1d(29413), Adra2a(25083), Adrb1(24925), Adrb2(24176), Drd1a(24316), Drd2(24318)

Gene Information

human ... ADRA1A(148), ADRA1B(147), ADRA1D(146), ADRA2A(150), ADRA2B(151), ADRA2C(152), ADRB1(153), ADRB2(154), ADRB3(155)
rat ... Adra1a(29412), Adra2a(25083), Adrb1(24925), Adrb2(24176), Drd1a(24316), Drd2(24318)

Gene Information

-

Gene Information

human ... ADRA1A(148), ADRA1B(147), ADRA1D(146), ADRA2A(150), ADRA2B(151), ADRA2C(152), ADRB1(153), ADRB2(154), ADRB3(155)


Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

1–4 of 4 Questions  
  1. How long is a solution of Product A7257, (-)-Norepinephrine, stable?

    1 answer
    1. A stock solution of 1 mg/ml or greater may be stored frozen in aliquots. Avoid freeze-thaw cycles. When the solution gets a pink cast, the product has degraded.

      Helpful?

  2. How should Product A7257, (-)-Norepinephrine, be dissolved?

    1 answer
    1. This product is soluble in 0.5M HCl at a concentration of 50mg/mL, yielding a clear faint yellow solution.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What are the differences between Product No. A7257, (-)-Norepinephrine and Product No. A0937, (±)-Norepinephrine (+)-bitartrate salt?

    1 answer
    1. Product No. A7257, is the free base form of the (-) isomer; it is not water soluble. Product No. A0937 is the bitartrate salt of the racemic (+/-) mixture; it is soluble in water at a concentration of 50 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters