Skip to Content
Merck
All Photos(1)

Key Documents

92698

Sigma-Aldrich

Ethylenediamine-N,N′-disuccinic acid trisodium salt solution

~35% in H2O

Synonym(s):

N,N′-Ethylenedi-(aspartic acid) trisodium salt, EDDS-Na3, Trisodium ethylenediamine-N,N′-disuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N2Na3O8
Molecular Weight:
358.19
Beilstein:
8177837
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

~35% in H2O

functional group

amine
carboxylic acid

SMILES string

OC(CC(C(O)=O)NCCNC(C(O)=O)CC(O)=O)=O.C

InChI key

ZTUSPXCGDRIRFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediamine-N,N′-disuccinic acid trisodium salt (EDDS) is an aminopolycarboxylic acid (APCA), which is used as a complexing agent. It forms stable water-soluble complexes with metal ions. EDDS has a better chelating property than EDTA. It can also be used as a flushing agent due to its eco-friendly characteristics and high biodegradability.

Application

EDDS (chelating agent) along with natural montmorillonite (NM) can be used in the photochemical degradation process of 17β-estradiol (E2). EDDS-NM complex can also be employed as an efficient photocatalysis system for wastewater treatment. Cu and Zn ions in the contaminated agricultural soil can be removed using EDDS via Cu and Zn complex formation.
Ethylenediamine-N,N′-disuccinic acid trisodium salt solution has been used as a chelate to study its effect on Eucalyptus camaldulensis tree under controlled deficit irrigation (CDI) condition.
powerful, readily biodegradable chelant

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelant-enhanced heavy metal uptake by Eucalyptus trees under controlled deficit irrigation.
Fine P, et al.
The Science of the Total Environment, 493, 995-1005 (2014)
Marco Race et al.
International journal of environmental research and public health, 15(3) (2018-03-23)
This paper presents the results of an experimental study aimed at investigating the effect of operative parameters on the efficiency of a soil flushing process, conducted on real contaminated soil containing high amounts of Cu and Zn. Soil flushing tests
Jing Li et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 11(12), 1880-1885 (2012-08-30)
In this work, the photochemical degradation process of 17β-estradiol (E2) in a suspension of natural montmorillonite (NM) has been studied. The addition of the iron complexing agent ethylenediamine-N,N'-disuccinic acid (EDDS) to the suspension was investigated and compared with the system
Daniel R McDougall et al.
Aquatic toxicology (Amsterdam, Netherlands), 228, 105645-105645 (2020-10-04)
The toxicity of heavy metals commonly impacts the survival of crustacean and bivalve larvae in hatchery culture, and this has led to the widespread use of EDTA to decrease this toxicity. Since EDTA has a very poor biodegradability leading to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service