Skip to Content
Merck
All Photos(1)

Key Documents

914665

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate

average Mn ~14,600

Synonym(s):

F-108 diacrylate, PEG-PPG-PEG, Pluronic diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H3O[C2H4O]x[C3H6O]y[C2H4O]zC3H3O2
UNSPSC Code:
12352112
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn ~14,600

color

off-white to yellow

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Poloxamers are nonionic, triblock copolymers containg a central hydrophobic chain of poly(propylene glycol) (PPG) and two terminal hydrophilic chains of poly(ethylene glycol) (PEG). Due to their amphiphilic structure, poloxamer solutions feature temperature dependent self-assembling and thermo-gelling behavior. Concentrated aqueous solutions of poloxamers are liquid at low temperature and form a gel at higher temperature, in a reversible process. This characteristic has allowed for these materials to be used as drug carriers for most routes of administration including oral, topical, intranasal, ocular, and parenteral. Acrylate-modified derivatives are most commonly used as thermosensitive hydrogels for drug delivery and 3D bioprinting applications. After casting or printing, the material can be crosslinked to preserve the hydrogel structure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-Assembly of Pluronic F127 Diacrylate in Ethylammonium Nitrate: Structure, Rheology, and Ionic Conductivity before and after Photo-Cross-Linking.
Carlos R L, et al.
Macromolecules, 49(14), 5179-5189 (2016)
Smart hydrogels for 3D bioprinting.
Shuai W, et al.
International Journal of Bioprinting, 1(1), 3-14 (2015)
Self-Assembly of Pluronic F127 Diacrylate in Ethylammonium Nitrate: Structure, Rheology, and Ionic Conductivity before and after Photo-Cross-Linking.
Lopez Barron C R, et al.
Macromolecules, 49(14), 5179-5189 (2016)
Application of thermoreversible pluoronic F-127 gels in Pharmaceutical formulations.
Escobar-Chavez J J, et al.
J. Pharm. Pharm. Sci., 9(3), 339-358 (2006)
Christopher J Hansen et al.
Advanced materials (Deerfield Beach, Fla.), 25(1), 96-102 (2012-10-31)
Microvascular multinozzle arrays are designed and fabricated for high-throughput printing of functional materials. Ink-flow uniformity within these multigeneration, bifurcating microchannel arrays is characterized by computer modeling and microscopic particle image velocimetry (micro-PIV) measurements. Both single and dual multinozzle printheads are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service