Skip to Content
Merck
All Photos(3)

Key Documents

901701

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate

average Mn ~12,500

Synonym(s):

PEG-PPG-PEG, Poloxamer F127 diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H3O[C2H4O]x[C3H6O]y[C2H4O]zC3H3O2
UNSPSC Code:
51171641
NACRES:
NA.23

form

powder

mol wt

average Mn ~12,500

color

white to faint yellow

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate (PEG-PPG-PEG) is a bifunctional acrylate-based amphiphilic triblock copolymer, which can be used in applications such as pharmaceutics, biomedical and personal care products. It can also be used in the synthesis of alternating block polyurethanes, which can be potentially used in drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterizations and biocompatibility of alternating block polyurethanes based on P3/4HB and PPG-PEG-PPG
Li G, et al.
Journal of Biomedical Materials Research Part A, 98(1), 88-99 (2011)
Photopolymerization of Pluronic F127 diacrylate: a colloid-templated polymerization
Di Biase M, et al.
Soft Matter, 7(10), 4928-4937 (2011)
Photopolymerization of Pluronic F127 diacrylate: a colloid-templated polymerization.
Manuela, D. B et al.
Soft Matter, 7, 4928-4937 (2011)
Fabrication of mesoporous silica hollow spheres using triblock copolymer PEG-PPG-PEG as template
Wang X, et al.
Journal of Non-Crystalline Solids, 356(18-19), 898-905 (2010)
Christopher J Hansen et al.
Advanced materials (Deerfield Beach, Fla.), 25(1), 96-102 (2012-10-31)
Microvascular multinozzle arrays are designed and fabricated for high-throughput printing of functional materials. Ink-flow uniformity within these multigeneration, bifurcating microchannel arrays is characterized by computer modeling and microscopic particle image velocimetry (micro-PIV) measurements. Both single and dual multinozzle printheads are

Related Content

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service