Skip to Content
Merck
All Photos(1)

Key Documents

435457

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)

average Mn ~4,400

Synonym(s):

Pluronic® L-121, PEG-PPG-PEG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.3 mmHg ( 20 °C)

form

viscous liquid

mol wt

average Mn ~4,400

composition

PEG, 30 wt. %

refractive index

n20/D 1.454

surface tension

33 dyn/cm, 25 °C, 0.1 wt. % in H2O

viscosity

1,200 cP(25 °C, Brookfield)(lit.)

transition temp

cloud point 14 °C (1 wt. % aqueous solution)
softening point 5 °C

density

1.006 g/mL at 25 °C

HLB

1.0 - 7.0

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Surface active material whose properties are dependent on the hydrophilic (EO)/hydrophobic (PO) ratio. High EO content gives better o/w stabilizers.

Physical form

Terminal primary dihydroxy-functional oligomer.

Legal Information

Pluronic is a registered trademark of BASF

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liyan Zhao et al.
Colloids and surfaces. B, Biointerfaces, 97, 101-108 (2012-05-23)
In this study, curcumin (Cur) loaded mixed micelles (Cur-PF), composed of Pluronic P123 (P123) and Pluronic F68 (F68), was prepared using the thin-film hydration method and evaluated in vitro. The preparation process was optimized with a central composite design (CCD).
Wei Xu et al.
Drug delivery, 19(4), 208-219 (2012-05-31)
The aim of this study was to construct novel targeting polymeric micelles. Folate-Poly (ethylenimine)-Pluronic copolymers were synthesized. A paclitaxel (PTX)-loaded mixed micelles consisting of Folate-Poly (ethylenimine)-Pluronic and Pluronic L121 copolymers have been developed. The mixed micelles showed nano-sized spherical morphology.
V K Zhalimov et al.
Biofizika, 57(2), 308-316 (2012-05-19)
It has been shown that sorption of most proteins with the molecular weight lower than 200 kDa from human blood plasma on the surface of perfluorocarbon emulsion, stabilized with proxanol 268, is mainly based on hydrophobic interaction, whereas sorption of
A Kiwilsza et al.
Journal of microencapsulation, 30(1), 21-27 (2012-06-09)
Mesoporous material SBA-15 of hexagonal structure was synthesised and its usefulness as a carrier for a poorly soluble drug--lacidipine (LA)--was tested. The source of silica was tetraethyl orthosilicate (TEOS) and the structure ordering agent was non-ionic surfactant Pluronic P123. SBA-15
Jianan Shen et al.
Biomaterials, 34(5), 1581-1590 (2012-11-14)
The metastasis of breast cancer is the leading cause of cancer death in women, and the lung is a common location of a secondary tumor that has metastasized from the primary source tumor. In this work, an attempt to simultaneously

Articles

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service