Skip to Content
Merck
All Photos(2)

Key Documents

C105600

Sigma-Aldrich

Cyclohexanethiol

97%

Synonym(s):

Cyclohexyl mercaptan, Hexahydrothiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11SH
CAS Number:
Molecular Weight:
116.22
Beilstein:
1236342
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4 (vs air)

Quality Level

vapor pressure

10.3 mmHg ( 37.7 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

158-160 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

SC1CCCCC1

InChI

1S/C6H12S/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

CMKBCTPCXZNQKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - Pensky-Martens closed cup

Flash Point(C)

43 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danyu Liu et al.
Nature communications, 12(1), 778-778 (2021-02-05)
Whole series of nanoparticles have now been reported, but probing the competing or coexisting effects in their synthesis and growth remains challenging. Here, we report a bi-nanocluster system comprising two ultra-small, atomically precise nanoclusters, AuAg24(SR)18- and Au2Ag41(SR)26(Dppm)2+ (SR = cyclohexyl mercaptan, Dppm
Xi Kang et al.
Chemical science, 10(37), 8685-8693 (2019-12-06)
Structural transformations between isomers of nanoclusters provide a platform to tune their properties and understand the fundamental science due to their intimate structure-property correlation. Herein, we demonstrate a reversible transformation between the face-centered cubic (FCC) and icosahedral isomers of Pt1Ag28
Goonay Yousefalizadeh et al.
Nanoscale, 13(1), 242-252 (2020-12-18)
Noble metal clusters have unique photophysical properties, especially as a new class of materials for multiphoton biomedical imaging. The previously studied Au25SR18 exhibits "giant" two-photon absorbance cross sections. Herein, we investigate the origins of the large two photon absorption for
Fei Liu et al.
Journal of biological engineering, 13, 21-21 (2019-03-20)
Metastasis-associated lung adenocarcinoma transcript 1 (MALAT1), a long non coding RNA (lncRNA) present in serum, is an important biomarker for detecting hepatocellular carcinoma (HCC). However, there are some shortcomings in current detection methods. So developing other novel MALAT1 detection methods is
Chaitali Singhal et al.
International journal of biological macromolecules, 98, 84-93 (2017-01-28)
An impedimetric genosensor was fabricated for detection of hepatitis C virus (HCV) genotype 1 in serum, based on hybridization of the probe with complementary target cDNA from sample. To achieve it, probe DNA complementary to HCVgene was immobilized on the

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service