Skip to Content
Merck
All Photos(2)

Key Documents

694924

Sigma-Aldrich

Tris[2-phenylpyridinato-C2,N]iridium(III)

sublimed grade

Synonym(s):

Ir(ppy)3, Iridium, tris[2-(2-pyridinyl-κN)phenyl-κC]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H24IrN3
CAS Number:
Molecular Weight:
654.78
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

form

powder

loss

0.5% loss on heating, 359°C (typical, TGA)

transition temp

transition temp 437.6 °C (typical, DSC)

λmax

282 nm

fluorescence

λex 305 nm; λem 507 nm in chloroform

Orbital energy

HOMO 5.6 eV 
LUMO 3 eV 

OLED Device Performance

ITO/HMPD/TAZ:Ir(ppy)3 (7%)/Alq3/Al:Li

  • Color: green
  • Max. Luminance: 4000 Cd/m2
  • Max. EQE: 15 %

ITO/NPD/CBP:Ir(ppy)3 (6%)/Alq3/Mg:Ag
  • Color: green
  • Max. Luminance: 100000 Cd/m2
  • Max. EQE: 8 %
  • Turn-On Voltage: 4.3 V

ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: green
  • Max. Luminance: 207839 Cd/m2
  • Max. EQE: 21.6 %
  • Turn-On Voltage: 2.1 V

ITO/TCTA/Ir(ppy)3/Bphen/LiF/Al
  • Color: green
  • Max. Luminance: 300000 Cd/m2
  • Max. EQE: 19 %
  • Turn-On Voltage: 2.6 V

SMILES string

c1ccc(nc1)-c2ccccc2[Ir](c3ccccc3-c4ccccn4)c5ccccc5-c6ccccn6

InChI

1S/3C11H8N.Ir/c3*1-2-6-10(7-3-1)11-8-4-5-9-12-11;/h3*1-6,8-9H;

InChI key

QKBWDYLFYVXTGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OLED triplet emitter (green).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-efficiency, low-voltage phosphorescent organic light-emitting diode devices with mixed host
Kondakova, M.; Pawlik, T.; Young, R.; et al.
Journal of Applied Physics, 104, 094501-094501 (2008)
Highly efficient, single-layer organic light-emitting devices based on a graded-composition emissive layer
Holmes, R. J.; et al.
Applied Physics Letters, 97, 083308-083308 (2010)
Very high-efficiency green organic light-emitting devices based on electrophosphorescence
Forrest, S. R.; et al.
Applied Physics Letters, 75, 4-4 (1999)
High-efficiency organic electrophosphorescent devices with tris(2-phenylpyridine)iridium doped into electron-transporting materials
Adachi, C.; ea al.
Applied Physics Letters, 77, 904-904 (2000)
A highly efficient universal bipolar host for blue, green, and red phosphorescent OLEDs.
Ho-Hsiu Chou et al.
Advanced materials (Deerfield Beach, Fla.), 22(22), 2468-2471 (2010-05-07)

Articles

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

See All

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service