Skip to Content
Merck
All Photos(1)

Key Documents

901799

Sigma-Aldrich

Fluorescent nanodiamond

Nitrogen vacancy ~3 ppm NV centers, 140 nm avg. part. size, amine functionalized, powder

Synonym(s):

Fluorescent nanoparticles, Nanodiamonds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

form

nanoparticles
powder

composition

Nitrogen vacancy, ~3 ppm NV centers

avg. part. size

140 nm

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescent nanodiamond (nitrogen vacancy ~3 ppm NV centers, 140 nm avg. part. size, amine-functionalized, powder), a biocompatible carbon-based nanomaterial, has interesting properties like excellent photostability, low toxic and facile surface functionalizability. It can be used for a variety of biological applications.

Application

Nanodiamonds (NDs) containing NV color centers exhibit fluorescence in red/near infra-red (NIR) region. The fluorescence in NDs originates from nitrogen vacancies and, unlike quantum dots and organic dyes, NDs do not photobleach. Fluorescent NDs find applications in background-free imaging, flow cytometry, super-resolution imaging etc. Our aminated fluorescent nanodiamonds are well suited for conjugation with activated biomolecules.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The long-term stability and biocompatibility of fluorescent nanodiamond as an in vivo contrast agent
Vaijayanthimala V, et al.
Biomaterials, 33(31), 7794-7802 (2012)
Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting.
Chang B, et al.
Advances in Functional Materials, 23, 5737-5737 (2013)
Yuen Yung Hui et al.
Optics express, 18(6), 5896-5905 (2010-04-15)
Dynamics of fluorescent diamond nanoparticles in HeLa cells has been studied with two-photon fluorescence correlation spectroscopy (FCS). Fluorescent nanodiamond (FND) is an excellent fluorescent probe for bioimaging application, but they are often trapped in endosomes after cellular uptake. The entrapment
Fluorescent nanodiamond as a probe for the intercellular transport of proteins in vivo
Kuo Y, et al.
Biomaterials, 34(33), 8352-8360 (2013)
Mass production and dynamic imaging of fluorescent nanodiamonds
Chang YR, et al.
Nature Nanotechnology, 3(5), 284-288 (2008)

Articles

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service