Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fluorescent nanodiamond
Synonym(s):
Diamond nanopowder, Nanodiamonds
Linear Formula:
C
Products
Brand
Sigma-Aldrich (11)
Physical Form
Chemical Composition
Quality Segment
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
798169
Fluorescent nanodiamond,
Nitrogen vacancy ~2.5 ppm NV centers, 70 nm avg. part. size, 1 mg/mL in deionized water
Expand
View Pricing
798088
Fluorescent nanodiamond,
Nitrogen vacancy ~3 ppm NV centers, 120 nm avg. part. size, 1 mg/mL in deionized water
Expand
View Pricing
900174
Fluorescent nanodiamond,
Nitrogen vacancy 3 ppm NV/particle, 100 nm avg. part. size (DLS), 1 mg/mL in deionized water, hydroxylated
Expand
View Pricing
900173
Fluorescent nanodiamond,
Nitrogen vacancy >60 NVN/particle, 70 nm avg. part. size (DLS), 1 mg/mL in deionized water
Expand
View Pricing
900171
Fluorescent nanodiamond,
Nitrogen vacancy ~3 ppm, 100 nm avg. part. size (APS), aminated powder
Expand
View Pricing
798134
Fluorescent nanodiamond,
Nitrogen vacancy ~3ppm NV centers, 100 nm avg. part. size, 1 mg/mL in deionized water
Expand
View Pricing
798142
Fluorescent nanodiamond,
Nitrogen vacancy ~1.5ppm NV centers, 100 nm avg. part. size, 1 mg/mL in deionized water
Expand
View Pricing
901799
Fluorescent nanodiamond,
Nitrogen vacancy ~3 ppm NV centers, 140 nm avg. part. size, amine functionalized, powder
Expand
View Pricing
901800
Fluorescent nanodiamond,
nitrogen vacancy ~3 ppm NV centers, 140 nm avg. part. size, carboxylic acid functionalized, PEG 3000 coated, 1 mg/mL in deionized water
Expand
View Pricing
798150
Fluorescent nanodiamond,
Nitrogen vacancy ~3ppm NV centers, 90 nm avg. part. size, 1 mg/mL in deionized water
Expand
View Pricing
901802
Fluorescent nanodiamond,
nitrogen vacancy ~3 ppm, 140 nm avg. part. size, amine functionalized, PEG 3000 coated, 1 mg/mL in deionized water
Expand
View Pricing