Skip to Content
Merck
All Photos(5)

Key Documents

901046

Sigma-Aldrich

Carbon nanotube forestvertically aligned on Si substrate

Drawable, DxL 10-40 nm× 1.0 mm ± 0.5 mm

Synonym(s):

CNT array, MWCNT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Product Name

Carbon nanotube array, multi-walled, drawable, vertically aligned on Si substrate

description

Form: 10 mmx20 mm Si substrate

Quality Level

Assay

98% (carbon as CNT)

diam. × L

10-40 nm × 1.0 mm ± 0.5 mm

SMILES string

[C]

InChI

1S/C

General description

Multi-walled carbon nanotubes (MWCNT) are vertically aligned on a Si substrate with a high density. Since all the MWCNTs are bundled by van der Walls force, CNT web is drawn easily by pinching an edge of the array out from the substrate. The web is very light weight and robust, and sometimes semi-transparent.
Crystal quality of CNT is examined by Raman scattering measurement. Intensity ratio of G-peak (1580 cm-1) and D-peak (1350 cm-1) represents quality of graphitization of carbon materials. G/D of our CNT is ~3, showing high crystal quality.

Preparation Note

Chemical Vapor Deposition (CVD) method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CNT arrays: Vertically aligned nanotubes grown via CVD; drawn into pure, aligned sheets for innovations.

CNT arrays: Vertically aligned nanotubes grown via CVD; drawn into pure, aligned sheets for innovations.

CNT arrays: Vertically aligned nanotubes grown via CVD; drawn into pure, aligned sheets for innovations.

CNT arrays: Vertically aligned nanotubes grown via CVD; drawn into pure, aligned sheets for innovations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service