773735
Carbon nanotube, single-walled
(6,5) chirality, ≥95% carbon basis (≥95% as carbon nanotubes), 0.78 nm average diameter
Synonym(s):
CHASM™, CNT, Signis® SG65i, SWCNT, SWNT, Single wall carbon nanotube
About This Item
Recommended Products
description
G/D Ratio: ≥20 (Raman 633 nm)
Median length: 1 μm
Assay
≥95% carbon basis (≥95% as carbon nanotubes)
form
powder (freeze-dried)
manufacturer/tradename
Signis® SG65i
surface area
≥700 m2/g
impurities
≤5 wt. % Moisture content
average diameter
0.78 nm
mp
3652-3697 °C (lit.)
density
1.7-1.9 g/cm3 at 25 °C (lit.)
bulk density
0.1 g/cm3
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Preparation Note
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The CoMoCAT® method of single-walled carbon nanotube (SWNT) synthesis yields high purity SWNTs with specific chiralities and narrow distributions of tube diameters.
The CoMoCAT® method of single-walled carbon nanotube (SWNT) synthesis yields high purity SWNTs with specific chiralities and narrow distributions of tube diameters.
The CoMoCAT® method of single-walled carbon nanotube (SWNT) synthesis yields high purity SWNTs with specific chiralities and narrow distributions of tube diameters.
The CoMoCAT® method of single-walled carbon nanotube (SWNT) synthesis yields high purity SWNTs with specific chiralities and narrow distributions of tube diameters.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service