Skip to Content
Merck
All Photos(3)

Key Documents

659258

Sigma-Aldrich

Carbon nanotube, multi-walled

>90% carbon basis, D × L 110-170 nm × 5-9 μm

Synonym(s):

MWCNT, MWNT, Multiwall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

>90% carbon basis

form

powder

composition

carbon, >90%

D × L

110-170 nm × 5-9 μm

mp

3652-3697 °C (lit.)

density

1.7 g/mL at 25 °C
~2.1 g/mL at 25 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Multi walled carbon nanotubes (MWNTs, CNTs) were prepared by chemical vapor deposition (CVD). In chemical vapor deposition (CVD), a volatile precursor undergoes thermal decomposition at elevated temperatures to form a solid deposit on a substrate. 1 The diameter of the tube ranges between 110-170nm and the length is between 5-9μm. The multiwalled nanotubes are stable in inert atmosphere upto a temperature of 3697oC.

Application

Carbon nanotube, multi-walled (MWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, biosensor, gas sensor, supercapacitor and as a filler that acts as a reinforcement to improve the mechanical property of composites.
Toxic and immunological effects of carbon nanotubes on human colorectal cancer cell line (Caco-2), the human breast adenocarcinoma cell line(MCF-7), Calu-3 cells from human lung adenocarcinoma and serous cells of proximal bronchial airways was investigated. These multiwalled carbon nanotubes were used to design a 3D, free standing porous carbon structure. The self-supporting structures could find potential uses in energy storage, supercapacitors, photovoltaic cells, field emission devices, sensors and high performance catalysts.Polystyrene/MWNTs nanocomposites have been fabricated and studied for its electrical and optical properties. Preparation of platinum/MWNTs/cobaltite nanocomposites has also been reported.

Preparation Note

Chemical Vapor Deposition (CVD) Method

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon nanotubes toxicology and effects on metabolism and immunological modification in vitro and in vivo.
Chiaretti M, et al.
Journal of Physics. Condensed Matter : An Institute of Physics Journal, 20(47) (2008)
Electrical and optical percolations of polystyrene latex?multiwalled carbon nanotube composites.
kara S, et al.
Journal of Colloid and Interface Science, 344(2), 395-401 (2010)
Ternary platinum-multiwall carbon nanotube-cobaltite composites for methanol electrooxidation.
Awasthi R and Singh RN
International Journal of Electrochemical Science, 6, 4775-4775 (2011)
Fabrication and characterization of three-dimensional macroscopic all-carbon scaffolds.
Carbon null
Non-functionalized multi-walled carbon nanotubes alter the paracellular permeability of human airway epithelial cells.
Rotoli BM, et al.
Toxicology Letters, 178(2), 95-102 (2008)

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Carbon nanotubes (CNTs) have received much attention since their discovery in 1991 by Sumio lijima1 due to their excellent mechanical, electrical, and optical properties.

A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.

Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service