Skip to Content
Merck
All Photos(2)

Key Documents

740411

Sigma-Aldrich

Potassium azide

≥99.9% trace metals basis

Synonym(s):

Azidopotassium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KN3
CAS Number:
Molecular Weight:
81.12
EC Number:
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.9% trace metals basis

form

solid

mp

~300 °C (decomposition)

functional group

azide

SMILES string

[K+].[N-]=[N+]=[N-]

InChI

1S/K.N3/c;1-3-2/q+1;-1

InChI key

TZLVRPLSVNESQC-UHFFFAOYSA-N

Related Categories

Application

Potassium azide can be used as a reagent to synthesize:
  • 3-O-(3-Azidoopropyl)estrone from 3-O-(3-methanesulfonyloxypropoxy)estrone via nucleophilic azidation in the presence of imidazolium catalyst.
  • Azido phenacyl derivatives from α-bromoacetophenone derivatives in the presence of tetrabutylammonium bromide as a catalyst.
  • 1-Azido-4-fluorobenzene from potassium (4-fluorophenyl)trifluoroborate in the presence of cupric triflate as a catalyst.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient method for synthesis of phenacyl derivatives under homogeneous phase transfer catalyst condition in aqueous media
Sayyahi S, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 22, 300-302 (2011)
tert-Alcohol-functionalized imidazolium ionic liquid: catalyst for mild nucleophilic substitution reactions at room temperature
Shinde S, et al.
Tetrahedron Letters, 50, 6654-6657 (2009)
A M Liashchuk et al.
Zhurnal mikrobiologii, epidemiologii, i immunobiologii, (4)(4), 65-68 (2006-09-01)
A highly purified TUL4-CBD chimeric protein was obtained by one stage purification method. TUL4-CBD protein consists of TUL4 Francisella tularensis mature peptide sequence, Gly-Ser spacer and cellulose binding domain (CBD) of Anaerocellum thermophilum. The TUL4-CBD protein was shown to induce
A M Egorov et al.
Biochimica et biophysica acta, 659(1), 141-149 (1981-05-14)
Modification of 12 arginine residues per molecule of formate dehydrogenase (formate : NAD+ oxidoreductase, EC 1.2.1.2.) from the methylotrophic bacterium, Achromobacter parvulus I, by 2,3-butanedione results in complete inactivation of the enzyme. Inactivation of the enzyme is reversible and proceeds
Copper-mediated functionalization of aryl trifluoroborates
Schimler S, et al.
Synlett, 27, 2279-2284 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service