Skip to Content
Merck
All Photos(1)

Documents

155071

Sigma-Aldrich

Azidotrimethylsilane

95%

Synonym(s):

Trimethylsilyl azide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiN3
CAS Number:
Molecular Weight:
115.21
Beilstein:
1903730
EC Number:
MDL number:
UNSPSC Code:
12352103
eCl@ss:
39100709
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

52-53 °C/175 mmHg (lit.)

density

0.868 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)N=[N+]=[N-]

InChI

1S/C3H9N3Si/c1-7(2,3)6-5-4/h1-3H3

InChI key

SEDZOYHHAIAQIW-UHFFFAOYSA-N

General description

Azidotrimethylsilane (TMSN3) is a a colorless and stable organosilane reagent. It shows very slow decomposition at high temperatures. It is a very commonly used azide source and has been used in the synthesis of aminotriazole ligands. Azidotrimethylsilane can be easily synthesised by adding chlorotrimethylsilane dropwise to a stirred solution of NaN3 in diethylene glycol dimethyl ether.

Application

Azidotrimethylsilane can be used as:
  • A nitrogen precursor to prepare GaN nanowire via metal-organic chemical vapor deposition method.
  • An electrolyte additive in Li-O2 batteries. The addition of TMSN3 results in the formation of robust solid electrolyte interphase.
  • An efficient reagent in the synthesis of tetrazoles, fullerenyl azide, and α-azido oximes.
  • A silylating agent in the O-trimethyl silylation of alcohols and phenols.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Covalent functionalization of epitaxial graphene by azidotrimethylsilane
Choi, Junghun, et al.
The Journal of Physical Chemistry C, 113(22), 9433-9435 (2009)
Synthesis, 106-106 (1988)
Azidotrimethylsilane
Li, B. L.
Synlett, 23(10), 1554-1555 (2012)
Journal of the American Chemical Society, 115, 9347-9347 (1993)
Well-defined poly (oxazoline)-b-poly (acrylate) amphiphilic copolymers: From synthesis by polymer-polymer coupling to self-organization in water
Guillerm, Brieuc, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 51(5), 1118-1128 (2013)

Articles

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service