Skip to Content
Merck
All Photos(1)

Key Documents

739405

Sigma-Aldrich

2-(1H-Imidazol-2-yl)pyridine

97%

Synonym(s):

2-(2′-Pyridyl)imidazole, 2-(2-Pyridyl)imidazole, 2-(Imidazol-2-yl)pyridine, 2-(Pyridin-2-yl)imidazole, Pyridine,2-imidazol-2-yl- (8Cl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

135-140 °C

SMILES string

c1ccc(nc1)-c2ncc[nH]2

InChI

1S/C8H7N3/c1-2-4-9-7(3-1)8-10-5-6-11-8/h1-6H,(H,10,11)

InChI key

SZXUTTGMFUSMCE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madalyn M McFarland et al.
Antimicrobial agents and chemotherapy, 60(12), 7017-7034 (2016-09-08)
Toxoplasma gondii is a ubiquitous apicomplexan parasite capable of infecting humans and other animals. Current treatment options for T. gondii infection are limited and most have drawbacks, including high toxicity and low tolerability. Additionally, no FDA-approved treatments are available for
Bashir Mustafa Mohamed et al.
Journal of nanobiotechnology, 9, 29-29 (2011-08-02)
Nanomaterials such as SiO2 nanoparticles (SiO2NP) are finding increasing applications in the biomedical and biotechnological fields such as disease diagnostics, imaging, drug delivery, food, cosmetics and biosensors development. Thus, a mechanistic and systematic evaluation of the potential biological and toxic
Nidia D Corona-Motolinia et al.
Molecules (Basel, Switzerland), 25(20) (2020-10-18)
Transition metal-based compounds have shown promising uses as therapeutic agents. Among their unique characteristics, these compounds are suitable for interaction with specific biological targets, making them important potential drugs to treat various diseases. Copper compounds, of which Casiopeinas® are an
Yonnie Wu et al.
BMC cell biology, 9, 26-26 (2008-05-22)
Mammalian cells transform into individual tubular straw cells naturally in tissues and in response to desiccation related stress in vitro. The transformation event is characterized by a dramatic cellular deformation process which includes: condensation of certain cellular materials into a
Jessica Wahsner et al.
Chemical reviews, 119(2), 957-1057 (2018-10-24)
Tens of millions of contrast-enhanced magnetic resonance imaging (MRI) exams are performed annually around the world. The contrast agents, which improve diagnostic accuracy, are almost exclusively small, hydrophilic gadolinium(III) based chelates. In recent years concerns have arisen surrounding the long-term

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service