Skip to Content
Merck
All Photos(1)

Documents

379433

Sigma-Aldrich

2,6-Bis(2-benzimidazolyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13N5
CAS Number:
Molecular Weight:
311.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

>300 °C (lit.)

SMILES string

c1cc(nc(c1)-c2nc3ccccc3[nH]2)-c4nc5ccccc5[nH]4

InChI

1S/C19H13N5/c1-2-7-13-12(6-1)21-18(22-13)16-10-5-11-17(20-16)19-23-14-8-3-4-9-15(14)24-19/h1-11H,(H,21,22)(H,23,24)

InChI key

JBKICBDXAZNSKA-UHFFFAOYSA-N

General description

2,6-Bis(2-benzimidazolyl)pyridine acts as tridentate nitrogen donor ligand and reacts with UO2Cl2·3THF to form complexes. It is reported as an efficient anion receptor providing chemical shift and optical modification based signals for the detection of acetate ions. 2,6-bis(2-benzimidazolyl)pyridine is a V-shaped ligand and its picrate Mn(II) complex has been prepared. 2,6-Bis(2-benzimidazolyl)pyridine is also reported as an optical sensor useful for the aniline recognition.

Application

2,6-Bis(2-benzimidazolyl)pyridine may be employed as a chemosensor for the detection of fluoride ions. It may be used as a sensor for the detection of toxic metabolites of benzene such as phenol, hydroquinone, resorcinol, catechol and p-benzoquinone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Utilization of 2, 6-bis (2-benzimidazolyl) pyridine to detect toxic benzene metabolites.
Chetia B and Iyer PK.
Tetrahedron Letters, 48(1), 47-50 (2007)
Liming Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(5), 1060-1063 (2009-10-17)
In this paper, an optical sensor, 2,6-bis(2-benzimidazolyl)pyridine, is reported to be highly selective towards aniline. A unique spectral response of 2,6-bis(2-benzimidazolyl)pyridine towards aniline is found, and a linear relationship between the sensor's emission intensity and the aniline's concentration is observed
Bolin Chetia et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 313-316 (2011-07-12)
2,6-Bis(2-benzimidazolyl)pyridine, bbp, a simple tridentate ligand, is employed as a receptor for the recognition of anions. The binding of anionic guest species with bbp can be studied in short duration using UV/vis spectroscopy, fluorescence spectroscopy and (1)H NMR techniques at
2, 6-Bis (2-benzimidazolyl) pyridine as a chemosensor for fluoride ions.
Chetia B and Iyer PK.
Tetrahedron Letters, 49(1), 94-97 (2008)
Huilu Wu et al.
European journal of medicinal chemistry, 45(11), 5324-5330 (2010-09-21)
A V-shaped ligand 2,6-bis(2-benzimidazolyl)pyridine (bbp) and its picrate Mn(II) complex have been synthesized and characterized by elemental analysis, molecular conductivities, (1)H NMR, IR, UV-vis spectra and X-ray single crystal diffraction. The crystal structure of [Mn(bbp)(2)](pic)(2)·2DMF is six-coordinated forming a distorted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service