Skip to Content
Merck
All Photos(1)

Documents

234672

Sigma-Aldrich

2,2′:6′,2′′-Terpyridine

98%

Synonym(s):

α,α′,α″-Tripyridyl, 2,6-Di(2-pyridyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3
CAS Number:
Molecular Weight:
233.27
Beilstein:
11199
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

98%

form

solid

mp

89-91 °C (lit.)

SMILES string

c1ccc(nc1)-c2cccc(n2)-c3ccccn3

InChI

1S/C15H11N3/c1-3-10-16-12(6-1)14-8-5-9-15(18-14)13-7-2-4-11-17-13/h1-11H

InChI key

DRGAZIDRYFYHIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2′:6′,2′′;-Terpyridine is a tridentate ligand that can be prepared in two steps, starting with 2-acetylpyridine. It coordinates with metal centers to form complexes. It is also utilized in the synthesis of chiral derivatives for asymmetric catalysis.

Application

Due to the presence of three near-coplanar nitrogen donor atoms, 2,2′:6′,2′′;-terpyridine may be used as a metal-binding domain to form metallo-supramolecular structures.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multinucleating 2, 2': 6', 2 ?-terpyridine ligands as building blocks for the assembly of co-ordination polymers and oligomers.
CargillaThompson AMW.
J. Chem. Soc., Dalton Trans., 24, 3467-3475 (1992)
New functional polymers and materials based on 2, 2': 6', 2 ?-terpyridine metal complexes.
Andres PR and Schubert US.
Advanced Materials, 16(13), 1043-1068 (2004)
2, 2': 6', 2'-Terpyridine.
Potts KT, et al.
Organic Syntheses, 189-189 (1986)
Complexes of the Ruthenium (II)-2, 2': 6', 2''-terpyridine Family. Effect of Electron-Accepting and-Donating Substituents on the Photophysical and Electrochemical Properties.
Maestri M, et al.
Inorganic Chemistry, 34(10), 2759-2767 (1955)
2, 2?: 6?, 2 ?-Terpyridines: From chemical obscurity to common supramolecular motifs
Constable EC
Chemical Society Reviews, 36(2), 246-253 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service