738883
1-Methyl-2-azaadamantane-N-oxyl
97%
Synonym(s):
1-Me-AZADO
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H16NO
CAS Number:
Molecular Weight:
166.24
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
87-93 °C
greener alternative category
storage temp.
2-8°C
SMILES string
CC12CC3CC(CC(C3)N1[O])C2
InChI
1S/C10H16NO/c1-10-5-7-2-8(6-10)4-9(3-7)11(10)12/h7-9H,2-6H2,1H3
InChI key
IBNXYCCLPCGKDM-UHFFFAOYSA-N
General description
1-Methyl-2-azaadamantane-N-oxyl (1-Me-AZADO), a sterically less hindered nitroxyl radical, is widely used as catalyst. It is chemically stable and exhibits superior catalytic performance.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.
Application
1-Methyl-2-azaadamantane-N-oxyl (1-Me-AZADO) may be employed as catalyst for the aerobic oxidation of alcohols and sterically hindered alcohols.
Catalytic oxidant for greener oxidation of alcohols under aerobic, solvent-free conditions. Recoverable and recyclable.
Solvent free aerobic oxidation of alcohols with 1-methyl-2-azaadamantane N-oxyl as a recyclable catalyst through phase separation
Solvent free aerobic oxidation of alcohols with 1-methyl-2-azaadamantane N-oxyl as a recyclable catalyst through phase separation
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Efficient oxidation of alcohols electrochemically mediated by azabicyclo-N-oxyls.
Demizu Y, et al.
Tetrahedron Letters, 49(1), 48-52 (2008)
Solvent free aerobic oxidation of alcohols with 1-methyl-2-azaadamantane N-oxyl as a recyclable catalyst through phase separation.
Kuang Y, et al.
Green Chemistry, 13(7), 1659-1663 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service