Skip to Content
Merck
All Photos(3)

Key Documents

471712

Sigma-Aldrich

tert-Butanol

anhydrous, ≥99.5%

Synonym(s):

2-Methyl-2-propanol, tert-Butyl alcohol, Trimethyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
Beilstein:
906698
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
83 °C (lit.)
vapor pressure:
31 mmHg ( 20 °C)
44 mmHg ( 26 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

2.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)
44 mmHg ( 26 °C)

Assay

≥99.5%

form

solid or liquid

autoignition temp.

896 °F

expl. lim.

8 %

impurities

<0.005% water

evapn. residue

<0.0003%

refractive index

n20/D 1.387 (lit.)

pH

7 (20 °C)

bp

83 °C (lit.)

mp

23-26 °C (lit.)

solubility

water: miscible

density

0.775 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)O

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3

InChI key

DKGAVHZHDRPRBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butanol (tBA) is a tertiary alcohol. It undergoes dehydration by reactive distillation in the presence of solid acid catalyst to form isobutylene with high selectivity.[1] Lipase-mediated alcoholysis of soybean oil deodorizer distillate (SODD) has been conducted in tBA (as reaction medium) to generate biodiesel.[2] Formation of 2-tert-butyl-p-cresol (TBC), 2,6-di-tert-butyl-p-cresol (DTBC) and tert-butyl-p-tolyl ether has been reported when p-cresol undergoes butylation by tBA in the presence of 12-tungstophosphoric acid supported on zirconia (TPA/ZrO2) catalyst.[3] The interaction behavior of tBA on the Si (001)-(2×1) surface has been assessed by using the ab-initio density functional theory (DFT) based on pseudopotential approach.[4]

Application


  • Synthesis of Ribose - Oleic Acid Esters in the Presence- and Absence of Candida antarctica Lipase B.: This study investigates the synthesis of ribose-oleic acid esters using tert-Butanol as a solvent, both with and without the presence of Candida antarctica Lipase B. The research explores the efficiency and effectiveness of the enzyme-catalyzed process, highlighting the impact of tert-Butanol on the synthesis reaction. The findings contribute to the understanding of esterification processes in analytical chemistry and have potential applications in developing new synthetic methods (Çetinkaya et al., 2020).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atomic and electronic properties of tert-butanol on the Si (001)-(2?1) surface.
Kaderoglu C, et al.
European Physical Journal B: Condensed Matter and Complex Systems, 76(3), 359-363 (2010)
Lipase-catalyzed biodiesel production from soybean oil deodorizer distillate with absorbent present in tert-butanol system.
Wang L, et al.
Journal of Molecular Catalysis. B, Enzymatic, 43(1), 29-32 (2006)
Tert-butanol dehydration to isobutylene via reactive distillation.
Knifton JF, et al.
Catalysis Letters, 73(1), 55-57 (2001)
Alkylation of p-cresol with tert-butanol catalyzed by heteropoly acid supported on zirconia catalyst.
Devassy BM, et al.
J. Mol. Catal. A: Chem., 210(1), 125-130 (2004)
Zaida Lentini et al.
Frontiers in plant science, 11, 522-522 (2020-05-28)
Embryo rescue (ER) in cassava breeding has several relevant applications, from the recovery of broad crosses to the recovery of seeds from the standard pollination program. Cassava fruit setting may drop from 100%, during the 1st week after pollination, to

Articles

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. is it packed in a glass bottle with AcroSeal cap?

    1 answer
    1. This product is packaged in a 100ml amber glass bottle with a SureSeal cap with septum and a 33mm red polypropylene cap with a PTFE liner. The AcroSeal system is from another manufacturer.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    0 answers

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service