Skip to Content
Merck
All Photos(3)

Key Documents

722987

Sigma-Aldrich

2-Cyano-2-propyl benzodithioate

>97% (HPLC)

Synonym(s):

2-Cyanopropan-2-yl benzodithioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NS2
CAS Number:
Molecular Weight:
221.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

>97% (HPLC)

form

solid or liquid

refractive index

n20/D 1.621

mp

28-31 °C

density

1.146 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(SC(=S)c1ccccc1)C#N

InChI

1S/C11H11NS2/c1-11(2,8-12)14-10(13)9-6-4-3-5-7-9/h3-7H,1-2H3

InChI key

IDSLBLWCPSAZBL-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of methacrylate and methacrylamide monomers. Chain Transfer Agent (CTA)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohan Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 34(27), 7998-8006 (2018-06-28)
Amphiphilic poly( N-(2-hydroxypropyl)methacrylamide) copolymers ( pHPMA) bearing cholesterol side groups in phosphate buffer saline self-assemble into nanoparticles (NPs) which can be used as tumor-targeted drug carriers. It was previously shown by us that human serum albumin (HSA) interacts weakly with
Ryuichi Nakatani et al.
ACS applied materials & interfaces, 9(37), 31266-31278 (2017-03-18)
In this study, a series of perpendicular lamellae-forming poly(polyhedral oligomeric silsesquioxane methacrylate-block-2,2,2-trifluoroethyl methacrylate)s (PMAPOSS-b-PTFEMAs) was developed based on the bottom-up concept of creating a simple yet effective material by tailoring the chemical properties and molecular composition of the material. The
Izumi Fukumoto et al.
PloS one, 8(11), e82540-e82540 (2013-12-07)
The immunostimulatory effects of the representative dental resin monomer 2-hydroxyethyl methacrylate (HEMA), a HEMA derivative that does not contain a double bond (2-hydroxyethyl isobutyrate, HEIB), and polymerized water-soluble oligomers of HEMA (PHEMA) were investigated. It is known that expression levels
Laure Gevaux et al.
Polymers, 11(2) (2019-04-10)
Fouling Release Coatings are marine antifouling coatings based on silicone elastomers. Contrary to commonly used biocide-based antifouling coatings, they do not release biocides into the marine environment, however, they suffer from poor antifouling efficacy during idle periods. To improve their
Thomas Swift et al.
Soft matter, 12(9), 2542-2549 (2016-01-30)
Fluorescence spectroscopy on a series of aqueous solutions of poly(acrylic acid) containing a luminescent label showed that polymers with molar mass, Mn < 16.5 kDa did not exhibit a pH responsive conformational change, which is typical of higher molar mass

Articles

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

See All

Protocols

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service