Skip to Content
Merck
All Photos(1)

Key Documents

11590

Sigma-Aldrich

4,4′-Azobis(4-cyanovaleric acid)

≥98.0% (T)

Synonym(s):

4,4′-Azobis(4-cyanopentanoic acid), ABCVA, ACVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCOCH2CH2C(CH3)(CN)N=NC(CH3)(CN)CH2CH2COOH
CAS Number:
Molecular Weight:
280.28
Beilstein:
1729856
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

solid

impurities

≤1% water

mp

118-125 °C (dec.) (lit.)

functional group

azo
carboxylic acid
nitrile

storage temp.

2-8°C

SMILES string

CC(CCC(O)=O)(\N=N\C(C)(CCC(O)=O)C#N)C#N

InChI

1S/C12H16N4O4/c1-11(7-13,5-3-9(17)18)15-16-12(2,8-14)6-4-10(19)20/h3-6H2,1-2H3,(H,17,18)(H,19,20)/b16-15+

InChI key

VFXXTYGQYWRHJP-FOCLMDBBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′-Azobis(4-cyanovaleric acid) has been used in the preparation of polystyrene particles by polymerizing styrene in ethyl alcohol.

Biochem/physiol Actions

4,4′-azobis(4-cyanovaleric acid) is an azo-initiator that induces lipid peroxidation of sunflower oil.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. D

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ray Chang et al.
Polymers, 10(10) (2019-04-10)
Temperature stimulus, easy modulation in comparison to other environmental stimuli, makes thermo-responsive nanocarriers popular in the applications of controlled drug release for cancer therapy. In this study, photosensitive sodium copper chlorophyllin (SCC) was incorporated into thermo-responsive polymeric nanogels consisted of
B Pekiner et al.
Biochemistry and molecular biology international, 33(6), 1159-1167 (1994-08-01)
Haemolysis induced by a free radical initiator (4,4'azobis-(4-cyanovaleric acid) has been studied with transfusion blood. Azobis cyanovaleric acid caused an increase in haemolysis. The effect of alpha- and gamma-tocopherol, 2,2,5,7,8 pentamethyl-6-chromanol and 2,2,7,8 tetramethyl-6-chromanol on the red blood cell haemolysis
Eric D Nelson et al.
Journal of pharmaceutical sciences, 95(7), 1527-1539 (2006-05-26)
AIBN and ACVA oxidative forced degradation models are examined for two drug molecules whose predominant oxidation chemistries arise from different reaction mechanisms (i.e., free radical vs. nucleophilic). Stress was conducted under a variety of initiator concentrations, and under ambient and
Dragana D Cetojević-Simin et al.
Journal of medicinal food, 13(2), 452-459 (2010-02-23)
In this study we investigated antioxidative and antiproliferative activity of different horsetail (Equisetum arvense L.) extracts. The antioxidative activity was measured by the electron spin resonance (ESR) spectroscopy-spin trapping method. The influence of different horsetail extracts during lipid peroxidation of
Uniform polymer particles by dispersion polymerization in alcohol.
Journal of Polymer Science Part A: Polymer Chemistry, 24(11), 2995-3007 (1986)

Articles

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service