Skip to Content
Merck
All Photos(1)

Key Documents

746304

Sigma-Aldrich

S,S-Dibenzyl trithiocarbonate

97%

Synonym(s):

Carbonic acid, trithio-, dibenzyl ester, Carbonotrithioic acid bis(phenylmethyl) ester, DBTTC, Dibenzyl carbonotrithioate, NSC 33081

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14S3
CAS Number:
Molecular Weight:
290.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

mp

29-33 °C

storage temp.

2-8°C

SMILES string

S=C(SCC1=CC=CC=C1)SCC2=CC=CC=C2

InChI

1S/C15H14S3/c16-15(17-11-13-7-3-1-4-8-13)18-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

LAKYXBYUROTWBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trithiocarbonate-based RAFT agent used as a bifunctional Chain Transfer Agent (CTA) in RAFT polymerization of block copolymers for drug delivery. Provides good control of polymerization of vinyl monomers, including styrene, methacrylate, and acrylamide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of stimuli-sensitive copolymers by RAFT polymerization: potential candidates as drug delivery systems
Tebaldi, M. L., Leal, D. A., Montoro, S. R., & Petzhold, C.
Journal of Materials Research (2014)
Rida Tajau et al.
Scientific reports, 10(1), 21704-21704 (2020-12-12)
Polymeric nanoparticles (NPs) are commonly used as nanocarriers for drug delivery, whereby their sizes can be altered for a more efficient delivery of therapeutic active agents with better efficacy. In this work, cross-linked copolymers acted as core-shell NPs from acrylated
Thiocarbonylthio compounds (SC (Z) SR) in free radical polymerization with reversible addition-fragmentation chain transfer (RAFT polymerization).
Chiefari, J., Mayadunne, R. T., Moad, C. L., Moad, G., Rizzardo, E., Postma, A., & Thang, S. H.
Macromolecules, 36(7), 2273-2283 (2003)
W Rusli et al.
Journal of colloid and interface science, 587, 393-401 (2020-12-29)
In preparing polymer capsules by vesicle templated emulsion polymerization, the initial size and morphology of the biomimetic vesicle template dictate the final size and morphology of the capsules. The presence of salts (NaCl, NaBr and LiCl) influences the size, dispersity
Kiwon Hwang et al.
Polymers, 12(4) (2020-04-23)
Tread is an important component that directly affects the performance of passenger car radial (PCR) tires. Styrene-butadiene rubber (SBR) is mainly used for tire tread and it includes solution styrene-butadiene rubber (SSBR) and emulsion styrene-butadiene rubber (ESBR). Although SSBR is

Articles

Low-cost, high-purity RAFT Agents are vital for industrial RAFT polymerization. We supply effective agents for this technology.

Low-cost, high-purity RAFT Agents are vital for industrial RAFT polymerization. We supply effective agents for this technology.

Low-cost, high-purity RAFT Agents are vital for industrial RAFT polymerization. We supply effective agents for this technology.

Low-cost, high-purity RAFT Agents are vital for industrial RAFT polymerization. We supply effective agents for this technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service