Skip to Content
Merck
All Photos(2)

Key Documents

71718

Sigma-Aldrich

Sodium L-lactate

≥99.0% (NT)

Synonym(s):

(S)-2-Hydroxypropionic acid sodium salt, L-Lactic acid sodium salt, Sarcolactic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NaO3
CAS Number:
Molecular Weight:
112.06
Beilstein:
4567087
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (NT)

form

solid

optical activity

[α]20/D −12.5±0.5°, c = 1% in H2O

optical purity

enantiomeric ratio: ≥97.0 (enzymatic)

feature

chiral

impurities

≤1% water

mp

163-165 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

[Na+].C[C@H](O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1

InChI key

NGSFWBMYFKHRBD-DKWTVANSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium L-lactate is the sodium salt of L-lactic acid and is widely utilized as a chiral building block in organic synthesis for the preparation of various chiral compounds. It can also be used as a mild reducing agent in synthetic chemistry applications.

Application

Sodium L-lactate can be used as a:
  • Reagent for the dissolution of calcium D-gluconate.[1]
  • Crystallizing agent in the synthesis of Sn-containing silicate catalysts. [2]
  • Chiral building block to prepare chiral pyrrolidine intermediate, which is further used to synthesize chiral pyrrolidinium ionic liquids.
  • Mild reducing agent in the selective reduction of CuO into Cu nanorods.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spontaneous supersaturation of calcium D-gluconate during isothermal dissolution of calcium L-lactate in aqueous sodium D-gluconate
Martina V et al,
Food & Function, 5, 85-91 (2014)
Clinical evaluation of sodium bicarbonate, sodium L-lactate, and sodium acetate for the treatment of acidosis in diarrheic calves.
Kasari TR
Journal of the American Veterinary Medical Association, 187(4), 392-397 (1985)
Jian-Hua Mao et al.
Microbiome, 8(1), 53-53 (2020-04-18)
Recent evidence has linked the gut microbiome to host behavior via the gut-brain axis [1-3]; however, the underlying mechanisms remain unexplored. Here, we determined the links between host genetics, the gut microbiome and memory using the genetically defined Collaborative Cross
Lucy Liu et al.
Cell metabolism, 26(5), 719-737 (2017-10-03)
Elevated reactive oxygen species (ROS) induce the formation of lipids in neurons that are transferred to glia, where they form lipid droplets (LDs). We show that glial and neuronal monocarboxylate transporters (MCTs), fatty acid transport proteins (FATPs), and apolipoproteins are
Rupali Vohra et al.
Molecular neurobiology, 55(12), 9108-9121 (2018-04-13)
Müller cells are pivotal in sustaining retinal ganglion cells, and an intact energy metabolism is essential for upholding Müller cell functions. The present study aimed to investigate the impact of lactate on Müller cell survival and function. Primary mice Müller

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service