Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C3H5NaO3
CAS Number:
Molecular Weight:
112.06
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
4567087
grade
pharmaceutical primary standard
API family
sodium lactate
manufacturer/tradename
USP
mp
163-165 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[Na+].C[C@H](O)C([O-])=O
InChI
1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1
InChI key
NGSFWBMYFKHRBD-DKWTVANSSA-M
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Sodium lactate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Lactated Ringer′s Injection
- Potassium Chloride in Lactated Ringer′s and Dextrose Injection
- Modified Lactated Ringer′s and Dextrose Injection
- Half-Strength Lactated Ringer′s and Dextrose Injection
- Lactated Ringer′s and Dextrose Injection
- Multiple Electrolytes and Dextrose Injection Type 3
- Multiple Electrolytes Injection Type 2
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Multiple Electrolytes Injection Type 2
United States Pharmacopeia, 41(4), 1595-1595 (2021)
Lactated Ringer's and Dextrose Injection
United States Pharmacopeia, 45(2) (2021)
Lactated Ringer's Injection
United States Pharmacopeia, 45(2) (2021)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1614308-200MG | 04061835345595 |