Skip to Content
Merck
All Photos(1)

Key Documents

295973

Sigma-Aldrich

N-Phenyl-bis(trifluoromethanesulfonimide)

99%

Synonym(s):

1,1,1-Trifluoro-N-phenyl-N-[(trifluoromethyl)sulfonyl]methanesulfonamide, N,N-Bis(trifluoromethylsulfonyl)aniline, N-Phenyl-trifluoromethanesulfonimide, NSC 240874, Phenyl triflimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(SO2CF3)2
CAS Number:
Molecular Weight:
357.25
Beilstein:
1269141
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

100-102 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)S(=O)(=O)N(c1ccccc1)S(=O)(=O)C(F)(F)F

InChI

1S/C8H5F6NO4S2/c9-7(10,11)20(16,17)15(6-4-2-1-3-5-6)21(18,19)8(12,13)14/h1-5H

InChI key

DIOHEXPTUTVCNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acts as a transparent strong electron-withdrawing p-type dopant in carbon nanotubes

Reactant for:
Synthesis of amphoteric alpha-boryl aldehydes
Enantioselective synthesis of core ring skeleton of leucosceptroids A-D
Stereoselective synthesis of monoamine reuptake inhibitor NS9544 acetate
Stereoselective sulfoxidation
Mild triflating reagent.
Used recently to convert a ketone to an enol triflate with KHMDS (324671) and thence an olefin with Pd(PPh3)4 (216666) and Bu3SnH (234788).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 47, 3485-3485 (1991)
José M Mejía-Oneto et al.
Organic letters, 8(15), 3275-3278 (2006-07-14)
[Structure: see text] A new strategy for the synthesis of (+/-)-aspidophytine has been developed and is based on a Rh(II)-catalyzed cyclization/dipolar cycloaddition sequence. The resulting [3+2]-cycloadduct undergoes an efficient Lewis acid mediated cascade that rapidly provides the complete skeleton of
Journal of the American Chemical Society, 112, 8578-8578 (1990)
Chemistry Letters (Jpn), 7, 1313-1313 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service