Skip to Content
Merck
All Photos(1)

Key Documents

697737

Sigma-Aldrich

Tris-(8-hydroxyquinoline)aluminum

sublimed grade, 99.995% trace metals basis

Synonym(s):

8-Hydroxyquinoline aluminum salt, Alq3, Aluminum 8-hydroxyquinolinate, Aluminum oxinate, Tris-(8-hydroxyquinolinato)aluminum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H18AlN3O3
CAS Number:
Molecular Weight:
459.43
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Assay

99.995% trace metals basis

form

solid

loss

0.5 wt. % loss on heating, 332°C (typical, TGA)

mp

>300 °C (lit.)
411 °C (DSC)

λmax

259 nm

fluorescence

λex 390 nm; λem 519 nm

Orbital energy

HOMO 5.8 eV 
LUMO 3.1 eV 

OLED Device Performance

ITO/MoO3/NPD/Alq3/BPhen/LiF/Al

  • Color: green
  • Max. Luminance: 20000 Cd/m2
  • Max. EQE: 1.2 %
  • Turn-On Voltage: 2.8 V

ITO/NPD/CBP:Ir(ppy)3/BCP/Alq3/Mg:Al
  • Color: green
  • Max. Luminance: 100000 Cd/m2
  • Max. EQE: 8 %
  • Turn-On Voltage: 4.3 V

ITO/NPD/TCTA/BCPO:Ir(piq)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: red
  • Max. Luminance: 24529 Cd/m2
  • Max. EQE: 17 %
  • Turn-On Voltage: 2.7 V

ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: green
  • Max. Luminance: 207839 Cd/m2
  • Max. EQE: 21.6 %
  • Turn-On Voltage: 2.1 V

ITO/m-MTDATA/NPD/TTPhPhB/Alq3/LiF/Al
  • Color: blue
  • Max. Luminance: 9100 Cd/m2
  • Max. EQE: 2.5 %
  • Turn-On Voltage: 3.2 V

SMILES string

O([Al](Oc1cccc2cccnc12)Oc3cccc4cccnc34)c5cccc6cccnc56

InChI

1S/3C9H7NO.Al/c3*11-8-5-1-3-7-4-2-6-10-9(7)8;/h3*1-6,11H;/q;;;+3/p-3

InChI key

TVIVIEFSHFOWTE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Tris(8-hydroxyquinoline)aluminum(III)(Alq3) is a key green-light emitting and electron transport material for OLEDs. A study reports the new crystalline phase d besides its, originally known, three other crystalline phases, α,β,γ. It exhibits excellent chemical stability and high luminescence efficiencies.

Application

Key green-light emitting and electron transport material for OLEDs.
Thin films of tris(8-hydroxyquinoline) aluminum (Alq3) can be used for organic electroluminescent devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of blue luminescent tris (8-hydroxyquinoline ) alumunium (Alq3)
Colle M, et al.
Advances in Functional Materials null
Molecular Orbital Study of the First Excited State of the OLED Material Tris(8-hydroxyquinoline)aluminum(III)
Halls MD and Schlegel HB
Chemistry of Materials, 2632-2640 null
Molecular Orbital Study of the First Excited State of the OLED Material Tris(8-hydroxyquinoline)aluminum(III)
Halls MD and Schlegel HB
Chemistry of Materials null
Electronic structure of tris (8-hydroxyquinoline) aluminum thin films in the pristine and reduced states.
Johansson J, et al.
J. Chem. Phys. , 1 null
Preparation and characterization of blue luminescent tris (8-hydroxyquinoline ) alumunium (Alq3)
Colle M, et al.
Advances in Functional Materials null

Articles

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service