Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tris-(8-hydroxyquinoline)aluminum
Synonym(s):
8-Hydroxyquinoline aluminum salt, Alq
3
, Aluminum 8-hydroxyquinolinate, Aluminum oxinate, Tris-(8-hydroxyquinolinato)aluminum
Empirical Formula (Hill Notation):
C
27
H
18
AlN
3
O
3
CAS Number:
2085-33-8
Molecular Weight:
459.43
EC Number:
218-227-0
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Sublimed grade (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
444561
Tris-(8-hydroxyquinoline)aluminum,
99.995% trace metals basis
Expand
View Pricing
697737
Tris-(8-hydroxyquinoline)aluminum,
sublimed grade, 99.995% trace metals basis
Expand
View Pricing
416282
Tris-(8-hydroxyquinoline)aluminum,
98% trace metals basis
Expand
View Pricing