Skip to Content
Merck
All Photos(1)

Key Documents

444561

Sigma-Aldrich

Tris-(8-hydroxyquinoline)aluminum

99.995% trace metals basis

Synonym(s):

8-Hydroxyquinoline aluminum salt, Alq3, Aluminum 8-hydroxyquinolinate, Aluminum oxinate, Tris-(8-hydroxyquinolinato)aluminum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H18AlN3O3
CAS Number:
Molecular Weight:
459.43
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

powder

mp

>300 °C (lit.)

Orbital energy

LUMO -3.3 eV 
LUMO 3.3 eV 

OLED Device Performance

ITO/MoO3/NPD/Alq3/BPhen/LiF/Al

  • Color: green
  • Max. Luminance: 20000 Cd/m2
  • Max. EQE: 1.2 %
  • Turn-On Voltage: 2.8 V

ITO/NPD/CBP:Ir(ppy)3/BCP/Alq3/Mg:Al
  • Color: green
  • Max. Luminance: 100000 Cd/m2
  • Max. EQE: 8 %
  • Turn-On Voltage: 4.3 V

ITO/NPD/TCTA/BCPO:Ir(piq)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: red
  • Max. Luminance: 24529 Cd/m2
  • Max. EQE: 17 %
  • Turn-On Voltage: 2.7 V

ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: green
  • Max. Luminance: 207839 Cd/m2
  • Max. EQE: 21.6 %
  • Turn-On Voltage: 2.1 V

ITO/m-MTDATA/NPD/TTPhPhB/Alq3/LiF/Al
  • Color: blue
  • Max. Luminance: 9100 Cd/m2
  • Max. EQE: 2.5 %
  • Turn-On Voltage: 3.2 V

SMILES string

O([Al](Oc1cccc2cccnc12)Oc3cccc4cccnc34)c5cccc6cccnc56

InChI

1S/3C9H7NO.Al/c3*11-8-5-1-3-7-4-2-6-10-9(7)8;/h3*1-6,11H;/q;;;+3/p-3

InChI key

TVIVIEFSHFOWTE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Alq3 crystals exist in two isomeric states, meridional and facial isomers. A third isomeric state ( γ) was studied by X-ray diffraction, the δ-Alq3 transforms into γ-form at ~180°C. Mechanism of the Alq3 phosphorescence has been studied.

Application

Key green-light emitting and electron transport material for OLEDs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triplet State Phosphorescence in Tris (8-hydroxyquinoline) Aluminum Light Emitting Diode Materials.
Perumal S, et al.
The Journal of Physical Chemistry C, 3446-3455 null
Green- and blue-emitting tris(8-hydroxyquinoline) aluminum(III) (Alq3) crystalline polymorphs: Preparation and application to organic light-emitting diodes.
Fukushim T and Kaji H
Organic Electronics null
Triplet State Phosphorescence in Tris (8-hydroxyquinoline) Aluminum Light Emitting Diode Materials.
Perumal S, et al.
The Journal of Physical Chemistry C null
Green- and blue-emitting tris(8-hydroxyquinoline) aluminum(III) (Alq3) crystalline polymorphs: Preparation and application to organic light-emitting diodes.
Fukushim T and Kaji H
Organic Electronics, 2985-2990 null
Highly Efficient Red Phosphorescent OLEDs based on Non-Conjugated Silicon-Cored Spirobifluorene Derivative Doped with Ir-Complexes
Lyu, Y.; Kwak, J.; Jeon, W.; et al.
Advances in Functional Materials, 19, 420-427 (2009)

Articles

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

See All

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service