695823
Di-1-adamantylphosphine
97%
Synonym(s):
Bis(adamant-1-yl)phosphine, Bis(tricyclo[3.3.1.13,7]dec-1-yl)phosphine
About This Item
Assay
97%
form
powder
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
181-185 °C
functional group
phosphine
SMILES string
C1C2CC3CC1CC(C2)(C3)PC45CC6CC(CC(C6)C4)C5
InChI
1S/C20H31P/c1-13-2-15-3-14(1)8-19(7-13,9-15)21-20-10-16-4-17(11-20)6-18(5-16)12-20/h13-18,21H,1-12H2
InChI key
RRRZOLBZYZWQBZ-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service