Skip to Content
Merck
All Photos(2)

Key Documents

287822

Sigma-Aldrich

Tri(o-tolyl)phosphine

97%

Synonym(s):

P(o-tol)3, Tris(o-tolyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4)3P
CAS Number:
Molecular Weight:
304.37
Beilstein:
661212
EC Number:
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: Cross Couplings
reaction type: Silylations
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

123-125 °C (lit.)

functional group

phosphine

SMILES string

Cc1ccccc1P(c2ccccc2C)c3ccccc3C

InChI

1S/C21H21P/c1-16-10-4-7-13-19(16)22(20-14-8-5-11-17(20)2)21-15-9-6-12-18(21)3/h4-15H,1-3H3

InChI key

COIOYMYWGDAQPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tri(o-tolyl)phosphine is a ligand used in the heck reaction and suzuki coupling of propargylic carbonates.

Application

Ligand used in a ruthenium-catalyzed direct amination of alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerio Cinà et al.
ChemPlusChem, 85(3), 391-398 (2020-03-03)
Hybrid nanostructures with switchable and reversible "blue-red-green" emission were efficiently synthesized. These nanostructures comprise polyhedral oligomeric silsesquioxanes (POSS) that behave as a nanocage that can be functionalized with terpyridine-based organic ligands, which can be easily complexed with europium (III) ions.
Majken Borup Thomsen et al.
Molecular imaging and biology, 22(5), 1290-1300 (2020-06-10)
Loss of neuronal synapse function is associated with a number of brain disorders. The [11C]UCB-J positron emission tomography (PET) tracer allows for in vivo examination of synaptic density, as it binds to synaptic vesicle glycoprotein 2A (SV2A) expressed in presynaptic
Tetrahedron Letters, 47, 8881-8881 (2006)
Yih-Chun Chen et al.
ChemSusChem, 11(18), 3225-3233 (2018-07-08)
We have demonstrated two novel donor-acceptor-donor (D-A-D) hole-transport material (HTM) with spiro[fluorene-9,9'-phenanthren-10'-one] as the core structure, which can be synthesized through a low-cost process in high yield. Compared to the incorporation of the conventional HTM of commonly used 2,2',7,7'-tetrakis[N,N-di(4-methoxyphenyl)amino]-9,9'-spirobifluorene (Spiro-OMeTAD)
Tri (o-tolyl) phosphine for highly efficient Suzuki coupling of propargylic carbonates with boronic acids
Junzhe X et al.
Chemical Communications (Cambridge, England), 54, 10451-10454 (2018)

Related Content

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service