Skip to Content
Merck
All Photos(1)

Key Documents

671800

Sigma-Aldrich

Di(1-adamantyl)benzylphosphine

95%

Synonym(s):

cataCXium® ABn

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37P
CAS Number:
Molecular Weight:
392.56
Beilstein:
9359745
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Asymmetric synthesis

functional group

phosphine

SMILES string

C1[C@H]2C[C@H]3C[C@@H]1C[C@@](C2)(C3)P(Cc4ccccc4)[C@@]56C[C@@H]7C[C@@H](C[C@@H](C7)C5)C6

InChI

1S/C27H37P/c1-2-4-19(5-3-1)18-28(26-12-20-6-21(13-26)8-22(7-20)14-26)27-15-23-9-24(16-27)11-25(10-23)17-27/h1-5,20-25H,6-18H2/t20-,21+,22-,23-,24+,25-,26-,27-

InChI key

ANIAFEJRWQDKDV-ANIFQACUSA-N

General description

sold in collaboration with Solvias AG

Application

Catalyst for:
  • Sonogashira coupling reaction
  • Ruthenium-catalyzed amination of secondary alcohols. with ammonia
  • Rhodium-catalyzed regioselective hydroformylation of alkenes
  • Reductive carbonylation
  • Suzuki coupling reaction

Legal Information

US7148176
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service