Skip to Content
Merck
All Photos(2)

Key Documents

658510

Sigma-Aldrich

Potassium sulfite

90%

Synonym(s):

dipotassiumsulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO3
CAS Number:
Molecular Weight:
158.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

SMILES string

[K+].[K+].[O-]S([O-])=O

InChI

1S/2K.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

BHZRJJOHZFYXTO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium sulfite (K2SO3) can be used as a catalyst in the oxidation of sulfur dioxide. It is also used as an anionic catalyst for the cyclotrimerization of aryl isocyanates to prepare heterocyclic symmetrical isocyanurates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Crich et al.
Journal of the American Chemical Society, 128(8), 2544-2545 (2006-02-24)
Alkylation of potassium selenosulfate with allylic halides gives Se-allyl seleno Bunte salts. On reaction with thiols at room temperature, these afford mixed dialkyl selenosulfides, which undergo 2,3-sigmatropic rearrangement with loss of selenium, either spontaneously or with assistance by triphenylphosphine, thereby
The reactivity of products of thermal interaction between ammonium meta-vanadate and potassium sulfite as catalysts for oxidation of sulfur dioxide
Fouda MFR, et al.
Applied Catalysis A: General, 223(1-2), 11-27 (2002)
Sulfate catalysed multicomponent cyclisation reaction of aryl isocyanates under green conditions
Dekamin, Mohammad G, et al.
J. Chem. Res. (M), 2005(3), 177-179 (2005)
Xinjian Shi et al.
Nature communications, 5, 4775-4775 (2014-09-03)
Tungsten trioxide/bismuth vanadate heterojunction is one of the best pairs for solar water splitting, but its photocurrent densities are insufficient. Here we investigate the advantages of using helical nanostructures in photoelectrochemical solar water splitting. A helical tungsten trioxide array is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service