Skip to Content
Merck
All Photos(1)

Key Documents

630586

Sigma-Aldrich

2′-Hydroxy-4′,6′-dimethoxyacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

80-84 °C (lit.)

functional group

ketone

SMILES string

COc1cc(O)c(C(C)=O)c(OC)c1

InChI

1S/C10H12O4/c1-6(11)10-8(12)4-7(13-2)5-9(10)14-3/h4-5,12H,1-3H3

InChI key

FBUBVLUPUDBFME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo dos Santos et al.
Archiv der Pharmazie, 339(5), 227-237 (2006-03-31)
Semi-empirical molecular orbital calculations at AM1 level were done with the aim to investigate the structure-activity relationships of antispasmodic activities of ten 2-(X-benzyloxy)-4,6-dimethoxyacetophenones with X = H, 4'-F, 4'-NO2, 4'-CH3, 4'-Cl, 3',4'-(CH3)2, 4'-OCH3, 4'-Br, 4'-OCH2C6H5, and 4'-C(CH3)3, against acetylcholine-induced contraction
Fungicide and fungiostatic effects of xanthoxyline.
V Cechinel Filho et al.
Journal of ethnopharmacology, 53(3), 171-173 (1996-09-01)
Paula Boeck et al.
Archiv der Pharmazie, 338(2-3), 87-95 (2005-03-31)
Chalcones and chalcone-like compounds, most of them new ones, prepared by base-catalyzed condensation of appropriate aldehydes and xanthoxyline, were tested for antifungal properties against a panel of yeasts, hialohyphomycetes as well as dermatophytes with the agar dilution assay. Results indicate
C Tringali et al.
Fitoterapia, 72(5), 538-543 (2001-06-29)
Analysis of the polar fractions of an EtOH extract obtained from the bark of the African medicinal plant Fagara macrophylla led to the isolation and identification of the alkaloids oblongine (6), tembetarine (7) and magnoflorine (8) and the flavonoid hesperidin
V Cechinel Filho et al.
Journal of pharmaceutical sciences, 84(4), 473-475 (1995-04-01)
The antispasmodic activity of several xanthoxyline derivatives against acetylcholine-induced contraction of the guinea pig ileum was evaluated in vitro. The acetophenones with two methoxyl groups, mainly in the 3,4 positions, exhibited potent antispasmodic activity. Modification of the hydroxyl group in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service