Skip to Content
Merck
All Photos(1)

Key Documents

10798

Sigma-Aldrich

Apigenin

≥95.0% (HPLC)

Synonym(s):

4′,5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
262620
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95.0% (HPLC)

impurities

~2% water

mp

>300 °C (lit.)

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-7,16-18H

InChI key

KZNIFHPLKGYRTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Apigenin is a 270 Da flavonoid, which belongs to the flavone group of glycosides. It is synthesized by a wide variety of plants including cereals, grains, tea and oranges.[1]

Application

Apigenin has been used as a calibration standard for the quantification of Hieracium pannosum Boiss,[2] Juniperus sp,[3] using high-performance liquid chromatography (HPLC) with Diode-Array Detection (DAD). It has been used to study effect on hormone secretion in the human adrenocortical H295R cells.[4]

Biochem/physiol Actions

A plant flavonoid that has been found to inhibit cell proliferation by arresting the cell cycle at the G2/M phase. Inhibition of growth through cell cycle arrest and induction of apoptosis appear to be related to induction of p53. Inhibits PMA-mediated tumor promotion by inhibiting protein kinase C and the resulting suppression of oncogene expression. It has also been reported to inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.
A plant flavonoid that has been found to inhibit cell proliferation by arresting the cell cycle at the G2/M phase. Inhibition of growth through cell cycle arrest and induction of apoptosis appear to be related to induction of p53. Inhibitory effects on tumor promotion may also be due to inhibition of kinase activity and the resulting suppression of oncogene expression. It has also been reported to inhibit topoisomerase I catalyzed DNA religation and enhance gap junctional intercellular communication.
Apigenin is a class II pharmaceutical drug and highly intestine permeable. It has antioxidant, anti-inflammatory and antitumorigenic functionality.[5] It is a potent inhibitor of the cluster of differentiation 38 (CD30) nucleotide adenine dinucleotide (NAD+) nucleosidase and could be used to suppress CD30 mediated metabolic syndrome.[6]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Inhibitor of human estrogen synthetase[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mixture effects of dietary flavonoids on steroid hormone synthesis in the human adrenocortical H295R cell line
Ohlsson A, et al.
Food And Chemical Toxicology, 48(11), 3194-3200 (2010)
Health functionality of apigenin: A review
Ali F, et al.
International journal of food properties, 20(6), 1197-1238 (2017)
Apigenin in cancer therapy: anti-cancer effects and mechanisms of action
Yan X, et al.
Cell & Bioscience, 7(1), 50-50 (2017)
Comparative Analysis of Chemical Profile, Antioxidant, In-vitro and In-vivo Antidiabetic Activities of Juniperus foetidissima Willd. and Juniperus sabina L.
Orhan N, et al.
The Iranian Journal of Pharmaceutical Research, 16(Suppl), 64-64 (2017)
Amy A Mrazek et al.
Pancreas, 48(5), 711-718 (2019-05-16)
Chronic pancreatitis is the consequence of multiple episodes of recurrent acute pancreatitis (RAP). We hypothesized that apigenin can minimize the sequelae of RAP by limiting acinar cells' proinflammatory signaling pathways. AR42J acinar cells were treated in vitro with transforming growth

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Questions

  1. what is the solubility and solubility of apigenin?

    1 answer
    1. This product is soluble at 10 mg/mL in 0.1M NaOH.

      Please see the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/PDF731713.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service