Skip to Content
Merck
All Photos(1)

Key Documents

D107808

Sigma-Aldrich

2′,6′-Dihydroxyacetophenone

97%

Synonym(s):

2-Acetyl-1,3-dihydroxybenzene, 2-Acetylresorcinol, DHAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3COCH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
1366061
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

156-158 °C (lit.)

SMILES string

CC(=O)c1c(O)cccc1O

InChI

1S/C8H8O3/c1-5(9)8-6(10)3-2-4-7(8)11/h2-4,10-11H,1H3

InChI key

YPTJKHVBDCRKNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junjie Hou et al.
Journal of biomedicine & biotechnology, 2010, 759690-759690 (2010-03-27)
Selecting an appropriate matrix solution is one of the most effective means of increasing the ionization efficiency of phosphopeptides in matrix-assisted laser-desorption/ionization time-of-flight mass spectrometry (MALDI-TOF-MS). In this study, we systematically assessed matrix combinations of 2, 6-dihydroxyacetophenone (DHAP) and diammonium
Ellen D Inutan et al.
Molecular & cellular proteomics : MCP, 10(2), M110-M110 (2010-09-22)
Laserspray ionization (LSI) mass spectrometry (MS) allows, for the first time, the analysis of proteins directly from tissue using high performance atmospheric pressure ionization mass spectrometers. Several abundant and numerous lower abundant protein ions with molecular masses up to ∼20,000
Gheorghita Zbancioc et al.
Ultrasonics sonochemistry, 19(3), 399-403 (2011-09-13)
A new, selective, straightforward and general method for preparation of highly functionalized coronands or spiro derivatives bearing 1,2-dihydroxyacetophenone unit, under conventional conditions and ultrasonic irradiation, is reported. The reaction setup involves only one step, acylation of an α-chloro-3,4-dihydroxyacetophenone with phthaloyl
MALDI mass spectrometry in the solution of some forensic problems.
R Seraglia et al.
Forensic science international, 146 Suppl, S83-S85 (2005-01-11)
Hsiou-Yu Ding et al.
Experimental dermatology, 20(9), 720-724 (2011-05-28)
Two natural acetophenone derivatives, 2,5-dihydroxyacetophenone (2,5-DHAP) and 2,6-DHAP, were purified from Cynanchum bungei and identified as murine tyrosinase inhibitors. Investigation into 2,5-DHAP showed it to be an uncompetitive inhibitor of murine tyrosinase (K(I) 0.28 mm). 2,5-DHAP strongly inhibited both melanogenesis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service