Skip to Content
Merck
All Photos(1)

Key Documents

172405

Sigma-Aldrich

Tetraethylene glycol dimethyl ether

≥99%, cross-linking reagent click chemistry

Synonym(s):

2,5,8,11,14-Pentaoxapentadecane, Bis[2-(2-methoxyethoxy)ethyl] ether, Dimethoxytetraethylene glycol, Dimethyltetraglycol, Tetraglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)4CH3
CAS Number:
Molecular Weight:
222.28
Beilstein:
1760005
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Tetraethylene glycol dimethyl ether, ≥99%

vapor density

7.7 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99%
≥99.0% (GC)

form

liquid

autoignition temp.

510 °F

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

impurities

≤1.0% Water (Karl Fischer)
≤50 ppm Trace peroxide (as H2O2)

refractive index

n20/D 1.432 (lit.)

bp

275-276 °C (lit.)

mp

−30 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

COCCOCCOCCOCCOC

InChI

1S/C10H22O5/c1-11-3-5-13-7-9-15-10-8-14-6-4-12-2/h3-10H2,1-2H3

InChI key

ZUHZGEOKBKGPSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetraethylene glycol dimethyl ether or tetraglyme is an organic aprotic colourless solvent with film forming ability.

Application

Tetraglyme coating was applied on glucose sensors, such as Nafion by RF to render it “non fouling” and enhancing linking of bio mimetic molecules to the surface. Tetraglyme helps in selective adsorption of proteins while promoting cell adhesion. Other general uses include as an electrolyte in lithium ion battery.

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongguang Zhang et al.
Nanoscale research letters, 9(1), 137-137 (2014-03-25)
A novel sulfur/graphene nanosheet (S/GNS) composite was prepared via a simple ball milling of sulfur with commercial multi-layer graphene nanosheet, followed by a heat treatment. High-resolution transmission and scanning electronic microscopy observations showed the formation of irregularly interlaced nanosheet-like structure
Luisa Mayorga Szott et al.
Journal of biomedical materials research. Part A, 96(1), 150-161 (2010-11-26)
Nonspecific protein adsorption, particularly fibrinogen (Fg), is thought to be an initiating step in the foreign body response (FBR) to biomaterials by promoting phagocyte attachment. In previous studies, we therefore prepared radiofrequency glow discharge (ethylene oxide)-like tetraglyme (CH(3)O(CH(2)CH(2)O)(4)CH(3)) coatings adsorbing
Min Zhang et al.
Journal of biomedical materials research. Part A, 89(3), 791-803 (2008-05-23)
Previous studies have showed that radio-frequency plasma deposited tetraglyme coatings greatly reduced fibrinogen adsorption (Gamma(Fg)) from highly diluted plasmas (0.1 and 1%) and subsequent platelet adhesion under static conditions. In this study, the protein resistant properties of tetraglyme were re-examined
Mingchao Shen et al.
Journal of biomaterials science. Polymer edition, 13(4), 367-390 (2002-08-06)
Polyethylene oxide (PEO) surfaces reduce non-specific protein and cell interactions with implanted biomaterials and may improve their biocompatibility. PEO-like polymerized tetraglyme surfaces were made by glow discharge plasma deposition onto fluorinated ethylene propylene copolymer (FEP) substrates and were shown to
Emily E Fenn et al.
Journal of the American Chemical Society, 131(15), 5530-5539 (2009-03-28)
Poly(ethylene) oxide (PEO) is a technologically important polymer with a wide range of applications including ion-exchange membranes, protein crystallization, and medical devices. PEO's versatility arises from its special interactions with water. Water molecules may form hydrogen-bond bridges between the ether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service