Skip to Content
Merck
All Photos(2)

Key Documents

110175

Sigma-Aldrich

Tetraethylene glycol

99%

Synonym(s):

Bis[2-(2-hydroxyethoxy)ethyl] ether, Tetra(ethylene glycol), Tetraglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)3CH2CH2OH
CAS Number:
Molecular Weight:
194.23
Beilstein:
1634320
EC Number:
MDL number:
UNSPSC Code:
12162002
eCl@ss:
39020714
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.7 (vs air)

Quality Level

vapor pressure

<0.01 mmHg

Assay

99%

mol wt

average Mn 200

refractive index

n20/D 1.459 (lit.)

bp

314 °C (lit.)

mp

−5.6 °C (lit.)

density

1.125 g/mL at 25 °C (lit.)

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

OCCOCCOCCOCCO

InChI

1S/C8H18O5/c9-1-3-11-5-7-13-8-6-12-4-2-10/h9-10H,1-8H2

InChI key

UWHCKJMYHZGTIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An oligomer of polyethylene glycol, tetra (ethylene glycol) (TEG) is a transparent, colorless, odorless, low volatility, hygroscopic liquid soluble in ethyl alcohol and very soluble in water.

Application

Used for the synthesis of tetraethylene glycol metha acrylate monomer.

Glow discharge plasma deposition of TEG renders surfaces resistant to protein adsorption and cellular attachment.

Caution

Do not confuse with glycofurol, which is also sometimes called tetraglycol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Nakamura et al.
Nanomaterials (Basel, Switzerland), 8(8) (2018-08-17)
Copper nitride particles have a low decomposition temperature, they absorb light, and are oxidation-resistant, making them potentially useful for the development of novel wiring inks for printing circuit boards by means of intense pulsed light (IPL) sintering at low-energy. Here
Andrea Paolella et al.
Nature communications, 8, 14643-14643 (2017-04-11)
Recently, intensive efforts are dedicated to convert and store the solar energy in a single device. Herein, dye-synthesized solar cell technology is combined with lithium-ion materials to investigate light-assisted battery charging. In particular we report the direct photo-oxidation of lithium
Nicholas M Negretti et al.
Scientific reports, 7(1), 15455-15455 (2017-11-15)
Bile plays an important role in digestion, absorption of fats, and the excretion of waste products, while concurrently providing a critical barrier against colonization by harmful bacteria. Previous studies have demonstrated that gut pathogens react to bile by adapting their
Rituparna Sengupta et al.
Nucleic acids research, 45(22), 12671-12680 (2017-10-17)
Significant, otherwise-unavailable information about mechanisms and transition states (TS) of protein folding and binding is obtained from solute effects on rate constants. Here we characterize TS for lac repressor(R)-lac operator(O) binding by analyzing effects of RO-stabilizing and RO-destabilizing solutes on
Guillaume Tonin et al.
Scientific reports, 7(1), 2755-2755 (2017-06-07)
Due to its high theoretical specific capacity, the lithium/sulfur battery is one of the most promising candidates for replacing current lithium-ion batteries. In this work, we investigate both chemical and morphological changes in the electrodes during cycling, by coupling operando

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service