Skip to Content
Merck
All Photos(1)

Key Documents

538582

Sigma-Aldrich

2,4-Dihydroxybenzoic acid hydrazide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OH)2C6H3CONHNH2
CAS Number:
Molecular Weight:
168.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

247-250 °C (lit.)

functional group

amine
hydrazine

SMILES string

NNC(=O)c1ccc(O)cc1O

InChI

1S/C7H8N2O3/c8-9-7(12)5-2-1-4(10)3-6(5)11/h1-3,10-11H,8H2,(H,9,12)

InChI key

GOPUHTXVZGIOHN-UHFFFAOYSA-N

General description

2,4-Dihydroxybenzoic acid hydrazide is a phenolic hydrazide. 2,4-Dihydroxybenzohydrazide can be prepared by employing ethyl 2,4-dihydroxybenzoate and hydrazine hydrate as starting reagent.

Application

2,4-Dihydroxybenzoic acid hydrazide may be used in the synthesis of:
  • 2,4-dihydroxy-N′-(4-methoxybenzylidene)benzohydrazide
  • 2,4-dihydroxy-N′-(2-hydroxy-4-methoxybenzylidene)benzohydrazide
  • (E)-2,4-dihydroxy-N′-(2-hydroxy-3-methoxy-5-nitrobenzylidene) benzohydrazide dihydrate
  • [N′-(5-chloro-2-oxidobenzyl-κO)-2,4-dihydroxybenzohydrazidato-κ2N′,O](methanol-κO)dioxidomolybdenum(VI)–4,4′-bipyridine
  • (E)-N′-(3,4-dimethoxybenzylidene)-2,4-dihydroxybenzohydrazide methanol solvate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 4-Dihydroxy-N'-(4-methoxybenzylidene) benzohydrazide.
Diao YP, et al.
Acta Crystallographica Section E, Structure Reports Online, 64(2), o470-o470 (2008)
(E)-N'-(3, 4-Dimethoxybenzylidene)-2, 4-dihydroxybenzohydrazide methanol solvate.
Zhang QL, et al.
Acta Crystallographica Section E, Structure Reports Online, 65(10), o2392-o2393 (2009)
[N'-(5-Chloro-2-oxidobenzyl-?O)-2, 4-dihydroxybenzohydrazidato-?2N',O](methanol-?O) dioxidomolybdenum (VI)?4, 4'-bipyridine (1/1).
Ngan NK, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(6), m747-m747 (2011)
2, 4-Dihydroxy-N'-(2-hydroxy-4-methoxybenzylidene) benzohydrazide.
Han YY and Zhao QR.
Acta Crystallographica Section E, Structure Reports Online, 66(5), o1041-o1041 (2010)
(E)-2, 4-Dihydroxy-N'-(2-hydroxy-3-methoxy-5-nitrobenzylidene) benzohydrazide dihydrate.
Han YY, et al.
Acta Crystallographica Section E, Structure Reports Online, 66(5), o1085-o1086 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service