Skip to Content
Merck
All Photos(3)

Documents

B13071

Sigma-Aldrich

Benzhydrazide

98%

Synonym(s):

Benzoic acid hydrazide, Benzoylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONHNH2
CAS Number:
Molecular Weight:
136.15
Beilstein:
471797
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

112-114 °C (lit.)

SMILES string

NNC(=O)c1ccccc1

InChI

1S/C7H8N2O/c8-9-7(10)6-4-2-1-3-5-6/h1-5H,8H2,(H,9,10)

InChI key

WARCRYXKINZHGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E Agostinelli et al.
The Biochemical journal, 324 ( Pt 2), 497-501 (1997-06-01)
A novel copper-depleted bovine serum amine oxidase (BSAO), in which about half the molecules contained the organic cofactor in the oxidized form, was prepared by adding a reductant in anaerobic conditions to the cyanide-reacted protein. The CuI-semiquinone formed in these
M Kobayashi et al.
Biochemical and biophysical research communications, 256(2), 415-418 (1999-03-18)
The amidase from Rhodococcus rhodochrous J1, which hydrolyses amide to acid and ammonia, was found to catalyze the synthesis of hydrazide using hydrazine as a substrate. This is the first report on the hydrazide synthesis through enzymatic reactions. The enzyme
Susan M Aitken et al.
The Biochemical journal, 375(Pt 3), 613-621 (2003-07-19)
Many compounds are oxidized by haem enzymes, such as peroxidases and cytochromes P450, to highly reactive intermediates that function as enzyme inactivators. To evaluate the potential of arylhydrazides as selective metabolically activated peroxidase inhibitors, the mechanism of HRPC (horseradish peroxidase
Venkataswamy Sorna et al.
Journal of medicinal chemistry, 56(23), 9496-9508 (2013-11-19)
Lysine specific demethylase 1 (LSD1) plays an important role in regulating histone lysine methylation at residues K4 and K9 on histone H3 and is an attractive therapeutic target in multiple malignancies. Here we report a structure-based virtual screen of a
Tetsuya Toya et al.
Bioorganic & medicinal chemistry, 10(4), 953-961 (2002-02-12)
We have investigated the biologically active conformation of the non-steroidal ecdysone agonist, 1-tert-butyl-1,2-dibenzoylhydrazine (RH-5849) by means of design, synthesis and conformational analysis of cyclic derivatives of RH-5849. Among the synthesized compounds, a 6-membered cyclic hydrazine bearing two benzoyl groups (5)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service