Skip to Content
Merck
All Photos(1)

Key Documents

T1388

Sigma-Aldrich

2-Thiophenecarboxylic acid hydrazide

≥98%

Synonym(s):

2-Thenoylhydrazine, 2-Thienoylhydrazine, 2-Thienyl hydrazide, 2-Thienylcarboxylic acid hydrazide, 2-Thiophenecarbohydrazonic acid, 2-Thiophenecarbonyl hydrazide, 2-Thiophenecarboxylic hydrazide, Thiophene-2-carbohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2OS
CAS Number:
Molecular Weight:
142.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

mp

136-139 °C (lit.)

SMILES string

NNC(=O)c1cccs1

InChI

1S/C5H6N2OS/c6-7-5(8)4-2-1-3-9-4/h1-3H,6H2,(H,7,8)

InChI key

SOGBOGBTIKMGFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bichitra K Biswal et al.
The Journal of biological chemistry, 280(18), 18202-18210 (2005-03-05)
Crystal structures of the RNA-dependent RNA polymerase genotype 2a of hepatitis C virus (HCV) from two crystal forms have been determined. Similar to the three-dimensional structures of HCV polymerase genotype 1b and other known polymerases, the structures of the HCV
W M Gross et al.
Journal of clinical microbiology, 21(4), 565-568 (1985-04-01)
In the context of a busy reference laboratory, radiometric selective inhibition tests were evaluated for rapid differentiation of Mycobacterium tuberculosis and Mycobacterium bovis and of the M. tuberculosis complex from other mycobacteria. p-Nitro-alpha-acetylamino-beta-hydroxypropiophenone at 5 micrograms and hydroxylamine hydrochloride at
Kee Woong Kwon et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(5), 6483-6496 (2019-02-13)
Bacillus Calmette-Guerin vaccine confers insufficient pulmonary protection against tuberculosis (TB), particularly the Mycobacterium tuberculosis (Mtb) Beijing strain infection. Identification of vaccine antigens (Ags) by considering Mtb genetic diversity is crucial for the development of improved TB vaccine. MTBK_20640, a new
B Körmendy
Acta veterinaria Hungarica, 43(4), 377-384 (1995-01-01)
The tuberculosis-free status of Hungarian cattle herds in the period between 1988 and the end of 1993 is evaluated. An epidemiological analysis of tuberculin tests, laboratory assays and allergic tests yielding positive results, summarized in three tables, is given with
Hongmin Kim et al.
Vaccines, 8(2) (2020-04-25)
Tuberculosis (TB) is one of the deadliest infectious diseases worldwide and is caused by Mycobacterium tuberculosis (Mtb). An effective vaccine to prevent TB is considered the most cost-effective measure for controlling this disease. Many different vaccine antigen (Ag) candidates, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service